missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ULTRAhyb™ Ultrasensitive Hybridization Buffer

Catalog No. am8669
Encompass
Change view
Click to view available options
Quantity:
125 mL
4 x 125 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM8669 4 x 125 mL
AM8670 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM8669 Supplier Invitrogen™ Supplier No. AM8669
Only null left

Proprietary blend of hybridization accelerators and blocking agents that greatly enhance the levels of hybridization

  • Signals that once took days to visualize are now apparent in hours
  • It is supplied in four bottles containing 125mL each
  • Increase the sensitivity of blot hybridization experiments
  • Increase random-primed DNA probe sensitivity up to 100x; RNA probe sensitivity 20x
  • Decrease hybridization time to 2 hours
  • Using standard hybridization buffers, only 1 to 5% of target molecules on a blot hybridizes to probe, making blots a relatively insensitive method for nucleic acid analysis (Vernier et al. (1996) Anal Biochem 235: 11-19); With ULTRAhyb Ultrasensitive Hybridization Buffer, the hybridization reaction approaches completion, so that as few as 10,000 molecules can be detected
  • Compatible with RNA and DNA probes labeled isotopically and nonisotopically in northerns, Southerns, and dot/slot blots
  • Contains 50% formamide and is compatible with positively charged membranes
  • Requires 10-100x less nonisotopic DNA probe than standard hybridization buffers, making it possible to perform many more hybridizations with a single labeling reaction

Applications:

DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store at 4°C.
Buffer Hybridization Buffer
Quantity 4 x 125 mL
Shipping Condition Room Temperature
Product Type Ultrasensitive Hybridization Buffer

For Research Use Only. Not for use in diagnostic procedures.