missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIzol™ Max™ Bacterial RNA Isolation Kit

Catalog No. 16096040
Encompass
Change view
Click to view available options
Quantity:
100 Preps
200 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16096040 200 Preps
16096020 100 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16096040 Supplier Invitrogen™ Supplier No. 16096040

The TRIzol™ Max™ Bacterial RNA Isolation Kit provides a simple and reliable method to improve the isolation of intact total RNA from gram-positive and gram-negative bacteria.

The TRIzol™ Max™ Bacterial RNA Isolation Kit provides a simple and reliable method to improve the isolation of intact total RNA from gram-positive and gram-negative bacteria. The kit utilizes both the Max™ Bacterial Enhancement Reagent and TRIzol™ Reagent to inactivate endogenous RNases and promote protein denaturing, improving RNA quality and integrity. Isolation of RNA takes about an hour.

Key Features of the TRIzol™ Max™ Bacterial RNA Isolation Kit:
• Formulated for the isolation of RNA from both Gram-positive and Gram-negative bacteria
• Uses two technologies for maximum RNA yield and purity
• Minimizes time required for RNA isolation by eliminating enzymatic and mechanical lytic steps

Two Products in One Kit
The TRIzol™ Max™ Bacterial RNA Isolation Kit combines Max™ Bacterial Enhancement Reagent with TRIzol™ Reagent. A 5-minute pre-treatment of bacteria with Max™ Bacterial Enhancement Reagent, containing chelating agents, detergent and buffer, denatures bacterial proteins and effectively deactivates endogenous RNases. Subsequent bacterial lysis by TRIzol™ Reagent generates a high-quality, minimally degraded RNA product.

Purified RNA is Ideal for Use with a Variety of Applications and Products
Bacterial RNA isolated using the TRIzol™ Max™ Bacterial RNA Isolation Kit is suitable for downstream applications such as real-time PCR (qPCR), RT-PCR, northern blotting, nuclease protection assays, cDNA synthesis, microarray analysis, and dot blot hybridization.

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Specifications

Content And Storage • 2 x 20 mL Max Bacterial Enhancement Reagent; room temperature
• 200 mL TRIzol Reagent; room temperature

Isolation Technology Organic Extraction
Purification Time 1 hr.
Elution Volume 50 μL
Sample Type Bacteria (Gram +/-)
Final Product Type Total RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling
Quantity 200 Preps
Shipping Condition Room Temperature
Starting Material Amount Cells: ≤108
Bacterial culture: 1.5 mL
Yield Gram-negative bacteria: >20 μg
Gram-positive bacteria: ≤3 μg
Show More Show Less
I'd like to isolate RNA from gram-positive bacteria. What products can you recommend for me? Is a modified lysis protocol needed?

For bacterial samples, you could use our TRIzol Max Bacterial RNA Isolation Kit (Cat. No. 16096040), that contains an extra reagent during homogenization to help with lysis of the bacterial cell walls. You could also use our PureLink RNA Mini Kit (Cat. No. 12183018A), for which you would need to prepare a lyosyme solution to lyse the cell wall.

The TRIzol Reagent protocol specifies centrifugation speeds of 12,000 x g for 10 mins for RNA precipitation, but my centrifuge only goes up to 5,000 x g. Can I still perform my experiment?

Yes, centrifugation speeds as low as 5,000 to 6,000 x g have been used, but the centrifugation time should be doubled to get the expected yields.

I accidentally added too much chloroform to my TRIzol Reagent reaction. What should I do?

If a large amount of chloroform was inadvertently added, you can add more TRIzol Reagent so that the ratio of 0.2 mL chloroform:1 mL TRIzol Reagent is maintained. If too much chloroform is added, this can drive the DNA, and eventually the protein, into the aqueous phase.

I inadvertently added isopropanol instead of chloroform. What should I do?

If isopropanol is inadvertently added at this step instead of chloroform, add more isopropanol to precipitate everything, then resuspend the pellet in TRIzol Reagent and use the protocol as specified. RNA yields will be compromised, but it may be possible to obtain a product in RT-PCR. A detailed protocol follows:

(1) Add more isopropanol so that the total volume of isopropanol equals the volume of TRIzol Reagent used. Spin at 7500 x g for 10 min at 4 degrees C.
(2) Pour off supernatant; allow relatively compacted pellet to air dry (doesn't have to be completely dry, just reduce the volume of ispropanol).
(3) Estimate the size of the pellet in microliters; add at least 15–20 volumes of TRIzol Reagent (e.g., for a 100 µL pellet, add at least 1.5 mL TRIzol Reagent).
(4) Break the pellet up well (you may have to use a hand-held homogenizer). Store the solution for 10–15 min. at room temperature; every 5 min or so, shake it by hand to make certain it is well dispersed.
(5) Proceed with the TRIzol Reagent protocol as written (i.e., add chloroform). Results will not be optimal, but it may be possible to get a product in RT-PCR.

What carrier do you recommend for isolation of RNA with TRIzol Reagent?

Glycogen can be included with your sample to improve yield, and remains with the RNA (glycogen is water soluble). Polyacrylamide can also be used as a carrier to precipitate small amounts of RNA.

At what points can I stop in the protocol for TRIzol Reagent?

There are a couple of possible stopping points in the RNA extraction protocol as shown below:

•After homogenization (before addition of chloroform), samples can be stored at 4 degrees C overnight or at –70 degrees C for at least 1 year.
•Homogenized samples can sit at room temperature for several hours before chloroform is added.
•Homogenized samples can be thawed and refrozen prior to use (necessary when researcher intends to do experiment, but then cannot continue).
• After RNA precipitation, during RNA wash, the RNA can be stored in 75% ethanol for at least 1 year at –20 degreesC, or at least 1 week at 4 degrees C.

For DNA extraction, the phenol phase and interphase can be stored at 4 degrees C overnight before DNA precipitation. Some customers have tried storing at 4 degrees C for a week and –20 degrees C for a year and still got good recovery.

Do you have recommendations for homogenizing my sample for RNA isolation with TRIzol Reagent?

You can homogenize your sample using a glass Teflon or power homogenzier (Polytron or Tekmar's Tissumizer) in a 1.5 microcentrifuge tube. Cultured cells do not have to be homogenized. Sonication will work to lyse cells in TRIzol reagent, but should only be performed if you do not plan on isolating DNA from your sample. Cells grown in monolayers can be lysed directly in the culture dish.

Can I use TRIzol Reagent to isolate RNA from tissue that was stored in RNAlater Reagent?

Yes, tissue stored in RNAlater Reagent can be used in the TRIzol Reagent. Remove the tissue from RNAlater Reagent, and immediately submerge in TRIzol solution.

What is the difference between your TRIzol Reagent and TRIzol LS Reagent?

TRIzol LS Reagent is a more concentrated formula, allowing for lower quantities of the reagent to be used relative to the sample. LS stands for liquid samples. TRIzol LS Reagent is formulated with a reduced volume of water to allow addition of a larger sample volume to a smaller volume of reagent. Therefore, when TRIzol Reagent and TRIzol LS Reagent are used in accordance with their respective protocols, they will perform identically and contain the same amount of chaotropic agents. The two reagents can be distinguisehd by color, where TRIzol LS Reagent is a darker, maroon red while TRIzol Reagent is lighter in color.

What does the TRI stand for in TRIzol Reagent?

The TRI stands for Total RNA Isolation. It also signifies the fact that this reagent can be used in the purification of RNA, DNA, and proteins from a single source.

What is the composition of your TRIzol Reagent?

TRIzol Reagent is a ready-to-use mixture of phenol, guanidine isothiocyanate, red dye, and other proprietary components.

Can I use TRIzol Reagent to isolate RNA from a sucrose gradient fraction?

Yes you can. Here is a reference to a paper, as well as a brief description of method from the paper.

Genes to Cells (2001) 6:121-129 (under the heading 'RNA isolation and RT-PCR')

TRIzol LS Reagent (LifeSciences) was used according to the manufacturer's instructions to extract total RNA from sucrose gradient fractions. Briefly, 250 mL of each fraction was added to 750 mL TRIzol LS Reagent and shaken vigorously for 15 s. After a 10-min incubation at room temperature, 150 mL chloroform was added, followed by vigorous shaking and brief incubation at room temperature. Samples were then spun at 14,000 g for 10 min in a tabletop microcentrifuge. Five micrograms of nuclease-free glycogen were added to 300 mL of the aqueous phase and nucleic acids were precipitated with the addition of an equal volume of 2-propanol. After centrifugation at 14,000 g for 30 min at room temperature, the pellet was washed once with 75% ethanol and resuspended in 20 mL of nuclease-free, sterile water. Five microlitres of total RNA were used as substrate for random-primed cDNA synthesis using Superscript II modified MMLV reverse transcriptase (Gibco/Life Sciences).

How does TRIzol Reagent work for RNA isolation?

TRIzol Reagent contains phenol and guanidine isothiocyanate, which allow for isolation of nucleic acids from proteins. Further partitioning of the nucleic acids occurs in a pH dependent manner. At pH 7.0 or higher, DNA and RNA partition into the aqueous phase. At an acidic pH, below 7.0, DNA is denatured and will move into the organic phase (interphase) yet the RNA remains in the aqueous phase.TRIzol Reagent has pH about 5.

If blood has been collected in EDTA, is the efficiency of the TRIzol Reagent RNA isolation procedure affected?

Blood collected with EDTA typically has the highest DNA contamination, blood collected with heparin typically has less than that collected with EDTA, and blood collected with citrate shows the least DNA contamination of the three. (Formulation for citrate solution: 3.8% (w/v) which is 3.8 g/100 mL of water. Use 0.5 mL for every 4.5 mL of blood. Rock gently back and forth after adding citrate solution to mix.) Adding 12 µL of 5 N acetic acid per milliliter of TRIzol Reagent may help, although there may still be a problem with DNA contamination. Using plasma or serum works best. The fresher the blood sample the better the RNA. Degraded RNA has been observed in blood that has been processed in as little as two hours after drawing.

After I add chloroform to the TRIzol Reagent mixture, the aqueous phase is pinkish in color. What is the reason behind this pinkish coloration and will it effect the quality of my RNA?

There are a couple of reasons that the aqueous phase may appear pinkish. It may be due to the tissue you are using. This is common with skin samples. It is assumed that there is fat in these samples, and the fat micelles “try to spin to the top of the tube” during the centrifugation but are unable to get there. In skin samples, the micelles pick up melanin pigment and cause the aqueous phase to appear colored. Fat micelles may also pick up pigment from the TRIzol Reagent itself, resulting in a pinkish color. If a sample is thought to contain fat, the sample homogenate in TRIzol Reagent may be centrifuged prior to addition of chloroform. The fat will appear as a clear layer at the top of the supernatant; this should be pipetted off and discarded.

In addition, if a sample contains a lot of blood, the aqueous phase may appear cloudy and/or yellowish (this may be due to iron in the hemoglobin coming out).

If the centrifuge used is not cold, the organic phase will be a deeper maroon color; some of this color may come into the aqueous phase and cause it to appear orange or yellow.

Alternatively, a pinkish aqueous phase may also be caused by over-dilution of the sample (i.e., the sample:TRIzol Reagent ratio which is greater than 1:10), as well as too much salt or protein in the sample. This can cause premature phase separation, which can be remedied by adding a bit more TRIzol Reagent to the sample. If the RNA is isolated from a pinkish aqueous phase, chances are that it will be contaminated with DNA. Although this should not be significant as TRIzol Reagent is formulated to prevent premature phase separation.

What is the difference between TRIzol and TRIzol LS Reagent?

TRIzol LS Reagent is a complete, ready-to use reagent for easy and simultaneous isolation of total RNA, DNA, and proteins from liquid samples. The reagent, a mono-phasic solution of phenol and guanidinium thiocyanate, is an improvement to the single-step RNA isolation method developed by Chomzcynski and Sacchi. TRIzol LS Reagent is similar to the original TRIzol Reagent in composition and results of use. TRIzol LS Reagent is designed for use with liquid samples such as blood and virus preparations in which large volumes of aqueous samples need to be processed, while TRIzol Reagent is designed for cell cultures or tissues. It is formulated to accommodate processing of more liquid sample per unit of reagent compared to the original formula.

The only difference between TRIzol Reagent and TRIzol LS Reagent is the concentration of components. TRIzol  LS Reagent is slightly more concentrated. The formula allows lower quantities of reagent to be used relative to a liquid sample. (TRIzol = 10:1 required, TRIzol LS = 3:1 required). If you buy TRIzol LS Reagent, but want to use it like TRIzol Reagent (on solid samples), there will probably be a decrease in yield vs. using regular TRIzol Reagent. TRIzol LS Reagent should NOT be used undiluted with solid samples. To dilute: take 750 µL TRIzol LS Reagent + (50 to 100 mg tissue + water to make 250 µL).

Do you have a reagent that will isolate RNA, DNA, and protein from the same sample?

Our TRIzol Reagent (Cat. No. 15596026) can isolate RNA, DNA, and protein from the same sample.

I want to extract DNA, and if possible, RNA from formalin-fixed specimens in paraffin blocks. Which product would work for me?

We recommend the RecoverAll Total Nucleic Acid Isolation Kit for FFPE (Cat. No. AM1975). This kit is optimized for isolation of both DNA and RNA from formalin or paraformalin-fixed, paraffin-embedded (FFPE).

Another option is TRIzol Reagent, but be sure to check the references listed below. Because paraffin is not soluble in TRIzol Reagent, paraffin-embedded tissues can be quick-heated to get the tissue out of the paraffin; any paraffin which remains will float to the top of the aqueous phase (and should be avoided). (If the slice is very thin, the whole slice can be added to the TRIzol Reagent, and hopefully, the tissue will be exposed to the reagent). Most of the references we surveyed do not provide quantitative data, because paraffin-embedded tissues are dramatically influenced by the action of nucleases prior to fixation and by the formalin fixation time.

The ability to detect specific housekeeping genes by PCR analysis with RNA or DNA extracted from these tissues is usually considered to be a positive result. We do not have a protocol per se, but we have spoken with customers who are doing this. We recommend deparaffinizing with xylene (or other organic), then grinding the sample very thoroughly in TRIzol Reagent (may require a Polytron); in most cases, you have to homogenize with vigor because the DNA is crosslinked and you have to get it free. Microcarrier is recommended since the RNA is crosslinked and fragmented. From this point, the standard isolation protocol can be used. They have found publications that show that the success of the isolation is dependent on how long the sample was fixed (there is an inverse relationship): Inoue, T., et. al., Pathology International (1996) Vol 46, Iss 12, pp. 997-1004.

How can I avoid genomic DNA contamination when isolating RNA with TRIzol Reagent?

If you will not need to isolate genomic DNA from the same sample and want to reduce the chance of gDNA contamination in your RNA, you should perform the optional centrifuge step mentioned in step 1 of the TRIzol Reagent manual prior to addition of chloroform.

After homogenizing your sample thoroughly in TRIzol Reagent, centrifuge the sample at 12,000 X g for 10 minutes at 4 degrees C. Genomic DNA, cellular membranes, and polysaccharides will form a pellet, and your RNA will be in the supernatant. Any lipids and fats in your sample may form a layer at the top of the solution as well. Remove the fat layer if necessary with a sterile tool and transfer the RNA supernatant to a new vial. Discard the DNA pellet.

Add chloroform to the RNA supernatant and proceed with the RNA isolation protocol.

To reduce gDNA contamination even more, you can treat your RNA after isolation with amplification grade DNase I. (Using non-amplification grade DNase I is not recommended, as it is not validated for absence of RNases and has been shown to degrade RNA samples in some cases.)

What can I do to increase the RNA yield using TRIzol Reagent from small amounts of starting samples?

Add 10 micrograms of RNase-free glycogen to less than 10 mg tissue or less than 1 X 10e6 suspension cells. Glycogen, unlike salmon sperm DNA carrier, can be added when TRIzol Reagent is added to sample.

If my tissue has a high content of proteoglycans and/or polysaccharides, what can I do to ensure that these compounds don't contaminate the RNA I get from my TRIzol Reagent purification?

Pellet polysaccharides (also pellets genomic DNA): Centrifuge following homogenization before adding chloroform at 12,000 X g at 4 degrees C for 10 min to pellet polysaccharides. In addition, you may need to do a high-salt isopropanol precipitation as follows. After collecting the aqueous phase, add 0.25 mL isopropanol and 0.25 mL of 0.8 M sodium citrate, 1.2 M NaCl per 1 ml TRIzol Reagent. Mix the solution, centrifuge, and proceed with isolation as described. This precipitates the RNA and maintains proteoglycans and polysaccharides in a soluble form. Samples known to have a high content of proteoglycans or polysaccharides include rat liver, rat aorta, and plants.

Can TRIzol Reagent be used to isolate RNA from small samples?

Yes. We have been successful in-house with 104 cells using 0.4 ml of TRIzol Reagent. When precipitating the RNA, add 5-10 µg of RNAse-free glycogen as a carrier to the aqueous phase.

Can purifications with TRIzol Reagent be scaled up?

Yes. Scale up linearly for tissues and suspension cells. For monolayer cells, scale up linearly based on the surface area of the plate, NOT the amount of cells. Use at least 1 ml of TRIzol Reagent for every 10 cm2 of surface area.

How can I increase the stability of the RNA isolated from TRIzol Reagent during storage?

RNA dissolved in deionized formamide can be stored at -70 degrees C for up to 1 year. To precipitate RNA from formamide, add NaCl to final concentration of 0.2 M followed by 4 volumes of ethanol. Incubate 3-5 min at room temperature and centrifuge at 10,000 x g for 5 min.

What type of tubes should I use with the TRIzol Reagent?

Use polypropylene tubes. Do not use tubes sensitive to phenol.

Can I use a table-top centrifuge when I use the TRIzol Reagent?

Yes, Use 2,600 X g at 4 degrees C for 60 min for phase separation and 30 min for RNA precipitation.

What will be the approximate aqueous volume when using TRIzol Reagent?

About 60% of the TRIzol Reagent volume becomes part of the aqueous phase after chloroform addition.

Why do I lose my DNA pellet after washing with 70% ethanol?

The DNA pellet from precipitation with isopropanol is easily dislodged when washing with 70% ethanol. It is best to remove the isopropanol supernatant and the ethanol wash by pipetting. Be careful not to shoot the washing buffer directly onto the pellet. Instead, allow the washing buffer to run over the pellet. Regardless of which manufacturer's miniprep kit you use, washing the pellet can be challenging because it is so small.

Can DNA purified with TRIzol Reagent be used in restriction digests or PCR reactions?

Yes. However, since the DNA is dissolved in 8 mM NaOH, the pH needs to be adjusted with HEPES. Please contact Technical Service for details. Depending on the restriction endonuclease, use 3 to 6 units per µg of DNA for between 3 hr and overnight. In a typical assay, 80 to 90% of the DNA is digestible.

Which Thermo Fisher Scientific products can be used to purify RNA from plants?

There are several Thermo Fisher Scientific products that can be used to purify RNA from plants. They are listed in alphabetical order:

Plant RNA Isolation Reagent.
--Plant RNA reagent is designed to isolate high yields of RNA from plant samples, even difficult ones such as conifer and plant seeds. This reagent does a good job of removing phenolics and starches.

TRIzol and TRIzol LS reagents.
--TRIzol Reagent is a good choice unless the tissue is very watery (such as fruit), in which case TRIzol LS Reagent would be recommended. When using TRIzol Reagent with plant tissue, we recommend that you homogenize 15 mg (total weight) leaf with ground glass homogenizer in 1 mL of TRIzol Reagent. Proceed with the rest of the protocol. In poinsettia, we have observed 33 µg RNA from 45 mg tissue (0.7 µg/mL) and from tobacco 76 µg RNA from 100 mg tissue (0.8 µg/mL).

Special considerations for RNA precipitation from tissue containing high amounts of proteoglycans and/or polysaccharides (as found in plants): Add to the aqueous phase 0.25 mL of isopropanol followed by 0.25 mL of a high-salt precipitation solution (0.8 M sodium citrate and 1.2 M NaCl) per 1 mL of TRIzol Reagent used for homogenization. Mix the resulting solution, centrifuge, and proceed with isolation as described in the protocol. The modified precipitation effectively precipitates RNA and maintains proteoglycans and polysaccharides in a soluble form. This procedure should ONLY be used if the sample is known to have a high content of proteoglycans and polysaccharides. To isolate pure RNA from plant material containing a very high level of polysaccharides, the modified precipitation should be combined with an additional centrifugation of the initial homogenate.

Special considerations for RNA precipitation from cotton leaf: Yields from 100 mg tissue were almost non-existent. We have noted that DNA yield from cotton leaf is 1/6 to 1/3 that of other plants, so there may be a correlation with RNA yield as well. Try using 3 to 5X more tissue and a microcarrier, as well as the pre-spin (before chloroform) and modified (high salt) precipitation. Also, some plants when homogenized in TRIzol Reagent will change the pH; it should be 5.5 (measuring with pH paper is accurate enough). If it is higher, adjust using glacial acetic acid.

What is the stability of TRIzol Reagent?

TRIzol Reagent has demonstrated stability of 12 months when stored at room temperature. However, we recommend storage at 2 to 8 degrees C for optimal performance.

Can I skip the phase separation step in your TRIzol-based RNA prep kits?

All our TRIzol-based RNA prep kits, including the Phasemaker Tubes (Cat. No. A33248) include a chloroform-based phase separation step that cannot be skipped.


For Research Use Only. Not for use in diagnostic procedures.