missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRI Reagent™ Solution

Catalog No. AM9738
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9738 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM9738 Supplier Invitrogen™ Supplier No. AM9738
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tri Reagent™ solution is a complete, ready-to-use reagent for the isolation of total RNA or the simultaneous isolation of RNA, DNA, and protein from a variety of biological samples.

Tri Reagent™ solution is a complete, ready-to-use reagent for the isolation of total RNA or the simultaneous isolation of RNA, DNA, and protein from a variety of biological samples. Key features:

  • Simple – a single reagent for isolation of DNA-free RNA, RNA-free DNA, and protein
  • Productive – obtain higher yields than with traditional guanidine thiocyanate or cesium chloride methods
  • Versatile – works with many sample sources, including human, plant, yeast, bacterial and viral samples
  • Flexible – easily scalable depending on starting material and desired yield

Using TRI Reagent™ solution
TRI Reagent™ starting material can be either fresh or frozen tissue, or cell samples of almost any size—the protocol is easily scalable. This RNA isolation method is well-recognized and referenced in numerous publications, a testament to its reliability.

How it works
TRI Reagent™ combines phenol and guanidine thiocyanate in a monophase solution to facilitate the immediate inhibition of RNase activity. Biological samples are homogenized or lysed in TRI Reagent™; the subsequent addition of bromochloropropane or chloroform results in the separation of the homogenate into aqueous and organic phases. RNA partitions to the aqueous phase, DNA to the interphase, and proteins to the organic phase. The RNA can then be precipitated from the aqueous phase with the addition of isopropanol. The entire protocol can be completed in approximately 1 hour.

Applications
The isolated RNA is suitable for any downstream application, including RT-PCR, amplification for array analysis, hybridization assays, or in vitro translation. PCR, Southern blotting, and cloning can be performed using DNA isolated with TRI Reagent™. Isolated proteins can be used in SDS-PAGE and western blotting.

Tri Reagent™ solution is incorporated into our RiboPure™ and MagMAX™-96 for Microarrays RNA isolation kits.

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage 100 mL TRI Reagent; 4°C

Isolation Technology Organic Extraction
Purification Time 1 hr.
Elution Volume Varies
Sample Type Bacteria, Cells, Plants, Tissue, Yeast
Final Product Type Total RNA, DNA, Protein
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern/Southern/Western blotting, in vitro translation, nuclease protection assays, nucleic acid labeling, hybridization, enzymatic assays, immunoprecipitation, gel shift assays, 2D gel electrophoresis
Quantity 100 mL
Shipping Condition Room Temperature
Starting Material Amount Bacteria: ≤107 cells
Cells: ≤106
Plant: ≤106 cells
Tissue: ≤100 mg
Yeast: ≤106 cells
Yield RNA: ≤15 μg (cells), ≤10 μg (tissue)
Show More Show Less
What type of sample can be used with TaqMan MicroRNA Assays?

One to ten nanograms of total RNA per 15 µL RT reaction is recommended for use with the TaqMan MicroRNA Assay. We have successfully detected miRNAs in as little as 25 pg of total RNA (~100 cells) in samples where the miRNA was highly abundant. The total RNA isolation procedure used must preserve small RNA species. We recommend the Invitrogen mirVana miRNA Isolation Kit (Cat. No. AM1560) or TRI reagent (Cat. No. AM9738).

For Research Use Only. Not for use in diagnostic procedures.