missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ THE RNA Storage Solution

Catalog No. AM7000
Encompass
Change view
Click to view available options
Quantity:
50 mL
10 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM7000 10 x 1 mL
AM7001 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM7000 Supplier Invitrogen™ Supplier No. AM7000

The Invitrogen RNA Storage Solution is a buffer that provides greater RNA stability than standard 0.1 mM EDTA or TE Buffer.

The Invitrogen RNA Storage Solution is a buffer that provides greater RNA stability than standard 0.1 mM EDTA or TE Buffer. This 1 mM sodium citrate, pH 6.4 ± 0.2 buffer is provided in one bottle containing 50 mL.

The last step in every RNA isolation protocol is to resuspend the purified RNA pellet. After painstakingly isolating the RNA, it is crucial that the pellet be suspended and stored in a safe, RNase-free solution. The RNA Storage Solution has two features that minimize base hydrolysis of RNA: a low pH, and sodium citrate, which is an efficient chelating agent. The RNA Storage Solution is compatible with all common RNA applications such as reverse transcription, in vitro transcription, northern analysis, and nuclease protection assays. The RNA Storage Solution is prepared from the purest reagents available, under scrupulously monitored conditions in a state-of-the-art manufacturing facility. It is then subjected to rigorous quality control procedures to guarantee their purity and performance.

Order Info

Shipping Condition: Room temperature

Specifications

Concentration 1 mM
Content And Storage Store at -20°C.
Shipped at ambient temperature.
Packaging Type Tube
Product Line Invitrogen
Product Type RNA Storage Reagent
Quantity 10 x 1 mL
Shipping Condition Room Temperature
Form Liquid
Can I use RNA Storage Solution (Cat. No. AM7000, AM7001) for resuspending dye-labeled RNA?

We have not tested RNA Storage Solution with dye-labeled RNA and therefore do not recommend using it for this application.

What is the concentration of RNA Storage Solution (Cat. No. AM7000, AM7001)?

The concentration of RNA Storage Solution is 1 mM sodium citrate, pH 6.4.

For Research Use Only. Not for use in diagnostic procedures.