missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ T Cell Leukemia (Jurkat) Total RNA

Catalog No. AM7858
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM7858 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM7858 Supplier Invitrogen™ Supplier No. AM7858

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The highest-quality RNA available from Invitrogen, it is DNase-treated and subjected to unsurpassed quality control standards. One tube containing 100μg is provided at a concentration of 1mg/mL.

The highest-quality RNA available from Invitrogen, it is DNase-treated and subjected to unsurpassed quality control standards. One tube containing 100μg is provided at a concentration of 1mg/mL. Intactness of mRNA, genomic DNA contamination, presence/absence of enzymatic inhibitors, and ribosomal RNA contamination in poly(A) preparations are all important aspects used to judge the quality of FirstChoice™ RNAs. The samples are processed using Invitrogen RNA isolation reagents to produce highly pure, intact RNA. A stringent DNase treatment is performed to ensure that the RNA is ready for use in any downstream application, including RT-PCR. Because it is impossible to remove every DNA molecule from an RNA sample, each preparation is tested to ensure that any residual DNA contamination is insignificant. The integrity of the RNA is verified by capillary electrophoresis using the Agilent™ 2100 bioanalyzer.

Order Info

Shipping Condition: Dry ice

Specifications

Cell Line Jurkat
Concentration 1 mg/mL
Content And Storage Store at or below -70°C. Avoid multiple freeze–thaw cycles. Aliquots of the product may be stored short-term at -20°C. DO NOT STORE IN FROST-FREE FREEZER.
Species Human
Sample Type Total RNA
Product Line Ambion
Quantity 100 μg
Sample Treatment DNase-Treated
Sample Source Cell Line
Shipping Condition Dry Ice
Do you offer isolated, purified RNA?

Yes, please visit this website (http://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/rna-extraction/rna-types/purified-rna.html) for our FirstChoice total RNA isolated from human tissues, as well as from E. coli and human cell lines.

For Research Use Only. Not for use in diagnostic procedures.