missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNaseAlert™ Lab Test Kit

Catalog No. AM1964
Encompass
Change view
Click to view available options
Quantity:
25 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM1964 25 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM1964 Supplier Invitrogen™ Supplier No. AM1964

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Invitrogen™ RNaseAlert™ Lab Test kit is a Patent-pending technology detects RNase activity in a convenient and sensitive assay that delivers results in real time.

Invitrogen™ RNaseAlert™ Lab Test kit is a Patent-pending technology detects RNase activity in a convenient and sensitive assay that delivers results in real time. Suitable for testing small sample numbers and can be used to ensure that solutions, tubes, tips, etc. are RNase-free; the kit contains sufficient reagents for 25 reactions.

  • Detects as little as 3.5 x 10-7 units (∼0.5 pg) of RNase A, with a fluorometer or by eye
  • Simple, straightforward 30 min. assay
  • Monitor RNase activity in column fractions during protein purification
  • Use with DNaseAlert™ to quantitatively detect both RNases and DNases in the same sample

Specifications

Format Lyophilized
Content And Storage Store Florescent Substrate, 10X RNaseAlert Lab Test Buffer, and RNase A at -20°C.

Store RnaseZap™ and nuclease-free water at room temperature.
Shipping Condition Dry Ice
Quantity 25 reactions
Reagent Type RNase Control Reagent
Product Line Ambion, RNaseAlert
What is the concentration of RNase A in the RNaseAlert Lab Test Kit (Cat. No. AM1964)?

The concentration of RNase A in the RNaseAlert Lab Test Kit is proprietary.

For Research Use Only. Not for use in diagnostic procedures.