missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNAqueous™-Micro Total RNA Isolation Kit

Catalog No. AM1931
Encompass
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM1931 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM1931 Supplier Invitrogen™ Supplier No. AM1931

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

50 Micro Filter Cartridge Assembly; 50 Micro Elution Tubes; 15mL Wash Solution 1 Concentrate; 28mL Wash Solution 2/3 concentrate; 25 Lysis Solution; 200μL LCM Additive; 55μL DNase 2U/μL; 600μL 10X DNase I Buffer; 150μL DNase Inactivation Reagent; 5mL Elution Solution

The RNAqueous™-Micro Kit is used for phenol-free total RNA isolation from micro-sized samples (e.g., from less than 500,000 cultured cells, from few as 10 laser capture microdissected cells, or from ≤10 mg of tissue) using a guanidinium-based lysis/denaturant and glass fiber filter separation technology.

The RNAqueous™-Micro Kit is used for phenol-free total RNA isolation from micro-sized samples (e.g., from less than 500,000 cultured cells, from few as 10 laser capture microdissected cells, or from ≤10 mg of tissue) using a guanidinium-based lysis/denaturant and glass fiber filter separation technology. DNA-free™ DNA removal reagents are included in the kit, making the isolated RNA especially suitable for RT-PCR. This kit provides:

• Highly concentrated RNA—for increased sensitivity in downstream assays
• Isolation of high-quality RNA from micro-scale samples
• Reagents/protocol for RNA recovery from LCM samples

How it works
Samples are solubilized in a powerful chaotropic solution, and RNA is purified on a glass fiber filter cartridge specifically designed for very small samples. A simple modification of the protocol allows recovery of both large and small RNA species including tRNAs, 5S rRNA, and microRNAs. The RNA is eluted in a minimal volume of 20 μL. Residual genomic DNA is efficiently removed using the DNA-free™ system included in the kit. The RNAqueous™-Micro Kit contains enough reagents for 50 RNA isolations of DNA-free RNA.

Not high-throughput compatible (Manual)

Applications
DNA and RNA Purification and Analysis, RNA Extraction, Total RNA Isolation, Total RNA from Animal Cells and Tissues

Specifications

Content And Storage • 50 Filter Cartridges with Collection Tubes; store at room temperature
• 50 Elution Tubes (1.5 mL); room temperature
• 15 mL Wash Solution 1 (concentrated); room temperature
• 28 mL Wash Solution 2/3 (concentrated); 4°C
• 25 mL Lysis Solution; 4°C
• 200 μL LCM Additive; -20°C
• 55 μL DNase; -20°C
• 600 μL 10X DNase I Buffer; -20°C
• 150 μL DNase Inactivation Reagent; -20°C
• 5 mL Elution Solution; room temperature
Isolation Technology Silica Spin Column
Purification Time 15 min. (After sample homogenization)
Elution Volume 10–20 μL
Sample Type Cells, Tissue, Laser Capture Microdissection (LCM)
Final Product Type Total RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Quantity 50 Preps
Shipping Condition Box 1: Dry Ice
Box 2: Room Temperature
Starting Material Amount Cells: ≤105
Tissue: ≤10 mg
Show More Show Less
What are the differences in the RNAqueous kits you offer?

RNaqueous kits are based on a rapid, filter-based RNA isolation system that does not require phenol, chloroform, or other toxic organic chemicals. The RNAqueous Total RNA Isolation Kit is designed for sample sizes of 10-75 mg of tissue or 10^6 to 10^7 cells, whereas the RNAqueous-Midi Total RNA Isolation Kit is designed for tissue samples of up to 0.5 g or 10^9 cells. The RNAqueous-Micro Total RNA Isolation Kit is optimized for the purification of total RNA from micro-sized samples such as 10-500,000 cultured cells, as few as 10 microdissected cells, or up to 10 mg tissue. The RNAqueous-96 Total RNA Isolation Kit utilizes a 96-well plate format for high-throughput RNA isolation from 100 to 2 x 10^6 cells or 0.1 to 1.5 mg of tissue. The RNAqueous-4PCR Total RNA Isolation Kit is designed for RNA isolation from small tissue samples (0.5-75 mg) and small- to medium-sized cultured cell samples (approximately 100-10^7 cells).

For Research Use Only. Not for use in diagnostic procedures.