missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNAqueous™-96 Total RNA Isolation Kit

Catalog No. AM1920
Encompass
Change view
Click to view available options
Quantity:
192 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM1920 192 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM1920 Supplier Invitrogen™ Supplier No. AM1920
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

The RNAqueous™-96 Kit is used for high-throughput, phenol-free total RNA isolation from 100–2,000,000 cells or 0.1–1.5 mg of tissue using a guanidinium-based lysis/denaturant and glass fiber filter separation technology.

The RNAqueous™-96 Kit is used for high-throughput, phenol-free total RNA isolation from 100–2,000,000 cells or 0.1–1.5 mg of tissue using a guanidinium-based lysis/denaturant and glass fiber filter separation technology. The kit contains sufficient reagents for 192 (2 x 96) RNA purifications. The RNAqueous™-96 Kit is ideal for the isolation of total RNA from multiple samples and very small samples.

Using the RNAqueous™-96 Kit
The procedure uses an RNA-binding glass-fiber filter and either vacuum filtration or centrifugation methods to provide high yields of intact RNA. An optional on-the-filter DNase treatment can be performed to ensure removal of genomic DNA for RT-PCR applications. The 96-well plates are compatible with most common 96-well plate vacuum manifolds and 96-well plate centrifuge rotors. The RNAqueous™-96 Kit provides consistently high yields of RNA from various amounts of starting material. The high quality of RNA isolated is illustrated (see figure).

Accessory product
The RNAqueous™-96 Automated Kit (Cat. No. AM1812) is the same product in a format suitable for automation.

Specifications

Content And Storage • 50 mL Nuclease-free Water; store at room temperature
• 50 mL Elution Solution; room temperature
• 2 Filter Plates; room temperature
• 2 Collection Plates; room temperature
• 2 Adhesive Plate Sealers; room temperature
• 2 Plate Lids; room temperature
• 50 mL Lysis/Binding Solution; 4°C
• 175 mL Wash Solution (concentrated); 4°C
• 53 mL Rebinding Mix; 4°C
• 12 mL DNase I Buffer; -20°C
• 1.25 mL DNase I; -20°C
Isolation Technology Silica Plate
Elution Volume ≥50 μL
Sample Type Cells, Tissue
Final Product Type Total RNA
High-throughput Compatibility High-throughput Compatible
For Use With (Equipment) MultiPROBE™ II HT
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Quantity 192 Preps
Shipping Condition Box 1: Dry Ice
Box 2: Room Temperature
Starting Material Amount Cells: ≤106
Tissue: ≤1.5 mg
Show More Show Less
What are the differences in the RNAqueous kits you offer?

RNaqueous kits are based on a rapid, filter-based RNA isolation system that does not require phenol, chloroform, or other toxic organic chemicals. The RNAqueous Total RNA Isolation Kit is designed for sample sizes of 10-75 mg of tissue or 10^6 to 10^7 cells, whereas the RNAqueous-Midi Total RNA Isolation Kit is designed for tissue samples of up to 0.5 g or 10^9 cells. The RNAqueous-Micro Total RNA Isolation Kit is optimized for the purification of total RNA from micro-sized samples such as 10-500,000 cultured cells, as few as 10 microdissected cells, or up to 10 mg tissue. The RNAqueous-96 Total RNA Isolation Kit utilizes a 96-well plate format for high-throughput RNA isolation from 100 to 2 x 10^6 cells or 0.1 to 1.5 mg of tissue. The RNAqueous-4PCR Total RNA Isolation Kit is designed for RNA isolation from small tissue samples (0.5-75 mg) and small- to medium-sized cultured cell samples (approximately 100-10^7 cells).

Can you please recommend the best manifold to use with the RNAqueous-96 Total RNA Isolation Kit (Cat. No. AM1920)?

The EveryPrep Universal Vacuum Manifold (Cat. No. K211101) is compatible with the RNAqueous-96 Total RNA Isolation Kit.

For Research Use Only. Not for use in diagnostic procedures.