missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNA Rapid Extraction Solution

Catalog No. AM9775
Encompass
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9775 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM9775 Supplier Invitrogen™ Supplier No. AM9775

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Invitrogen™ RNA Rapid Extraction Solution is to release RNA from the paper prior to RNA isolation. 10 mL is provided.

Invitrogen™ RNA Rapid Extraction Solution is to release RNA from the paper prior to RNA isolation. 10 mL is provided.

  • Rapid 5 min. procedure to release RNA from Whatman™ FTA™ cards
  • Ideal for high-throughput applications
  • Integrity of the RNA is unaffected

Ready for real-time PCR Disks punched from FTA™ Cards are simply dropped into RNA Rapid Extraction Solution and agitated for 5 minutes to release the sample for RNA isolation—while protecting the RNA from degradation. The RNA can then be purified from the RNA Rapid Extraction Solution using the MagMAX™-96 Viral RNA Isolation Kit (Cat. No. AM1836). The high-quality purified RNA is suitable for quantitative RT-PCR or other downstream RNA applications. Wha™an™ FTA™ Cards can be used with virtually any sample type including blood, cultured cells, buccal cells, plant material, bacteria, plasmids, microorganisms, solid tissue, viral particles, and more.

Specifications

Content And Storage Store at room temperature.
Isolation Technology Organic Extraction
Sample Type Plant Samples, Blood, Other Samples, Bacteria, Cells, Whatman FTA Cards Samples
Final Product Type Total RNA, Viral DNA/RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Equipment) MagMAX™-96 Viral RNA Isolation Kit
For Use With (Application) Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Northern Blotting
Includes Solution, without manual
Product Line Ambion
Quantity 10 mL
Shipping Condition Room Temperature
Target Viral DNA/RNA, Total RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.