missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RiboMinus™ Human/Mouse Transcriptome Isolation Kit

Catalog No. K155001
Encompass
Change view
Click to view available options
Quantity:
6 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K155001 6 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K155001 Supplier Invitrogen™ Supplier No. K155001
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Includes

Contains two modules; the RiboMinus Human/Mouse Probe Module and the Concentration Module. The RiboMinus Human/Mouse Probe Module contains the RiboMinus magnetic beads, the RiboMinus human/mouse probe, and binding buffer. The Concentration Module contains spin cartridges, wash tubes, recovery tubes, binding buffer, wash buffer, and RNase-free water.

The RiboMinus™ Human/Mouse Transcriptome Isolation Kit constitutes a novel purification system that enriches the whole spectrum of mRNA species by depleting large ribosomal RNA molecules in a total RNA sample.

The RiboMinus™ Human/Mouse Transcriptome Isolation Kit constitutes a novel purification system that enriches the whole spectrum of mRNA species by depleting large ribosomal RNA molecules in a total RNA sample. The kit removes >95% of 18S and 28S rRNA molecules from human or mouse total RNA without relying on current polyA+ purification methods. This increases the representation of mRNA species, including non-polyA-tailed, fragmented mRNA or other regulatory RNAs that cannot be purified via polyA-selection methods. Purified RNA is suitable for several downstream applications, including, library construction, RT-PCR, and microarray.

Not high-throughput compatible (Manual)

Specifications

Content And Storage The RiboMinus™ Human/Mouse Transcriptome Isolation Kit contains two modules: the RiboMinus™ Human/Mouse Probe Module and the Concentration Module

The RiboMinus™ Human/Mouse Probe Module contains the RiboMinus™ magnetic beads, the RiboMinus™ human/mouse probe, and binding buffer. Store at 4°C

The Concentration Module contains spin cartridges, wash tubes, recovery tubes, binding buffer, wash buffer, and RNase-free water. Store at room temperature.

All reagents are guaranteed stable for 6 months when properly stored.
Isolation Technology Magnetic Bead
Sample Type Total RNA
Final Product Type Transcriptome RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Microarray Analysis, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Northern Blotting, cDNA Library Construction
Product Line RiboMinus
Quantity 6 preps
Target Transcriptome RNA
System Type RiboMinus
Form Magnetic Bead
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.