Learn More
Invitrogen™ QuantumRNA™ Classic II 18S Internal Standard
Catalog No. am1717
Encompass
Change view
Click to view available options
Quantity:
100 Reactions
Catalog No. Quantity
AM1717 100 Reactions
1 options
Catalog No. AM1717 Supplier Invitrogen™ Supplier No. AM1717
The Invitrogen™ QuantumRNA™ 18S Internal Standard with Competimer™ technology uses specially modified Competimers of the same sequence as the normal 18S primers that cannot be extended.
The Invitrogen™ QuantumRNA™ 18S Internal Standard with Competimer™ technology uses specially modified Competimers of the same sequence as the normal 18S primers that cannot be extended. By adjusting the ratio of 18S Competimers to normal 18S mRNA primers, the signal for 18S mRNA can be attenuated to the level of very rare messages. They are used for quantitative end-point RT-PCR and include sufficient reagents for 100 reactions. Because of its invariant expression across tissues and treatments, 18S ribosomal RNA is an ideal internal control for quantitative RNA analysis. The Classic II 18S Internal Standards produce a 324 bp PCR product.
Accessory Products:
The QuantumRNA™ Classic 18S Internal Standard (Cat. No. AM1716) produce a 489 bp PCR product, and the QuantumRNA™ Universal 18S Internal Standard (Cat. No. AM1718) function across the broadest range of eukaryotes, including animals, plants, and many protozoa and produce a 315 bp PCR product.
Specifications
Product Line Ambion, QuantumRNA
Quantity 100 Reactions
Packaging Tube(s)
Includes 18S Primer Pair and 18S Competimer™s are stored at –20°C. Control Template RNA should be stored below –70°C. Nuclease-free water may be stored at any temperature.
For Research Use Only. Not for use in diagnostic procedures.