Learn More
Invitrogen™ PARIS™ Kit
Description
The Invitrogen™ PARIS™ system is for the isolation of both RNA and native protein from the same experimental sample. The kit also permits separation of nuclear and cytoplasmic fractions prior to RNA and/or protein isolation. The kit contains sufficient reagents for 50 purifications from1 to 75 mg of tissue or 100 to 107 cells each.
• Fast, simple procedure
• Isolate nuclear and cytoplasmic RNA and protein from cultured cells
• Isolate total RNA and protein from cultured cells or tissues
• No phenol extraction or alcohol precipitation
• Reduces time, cost, and variability between independent experimental samples
Isolation of high-quality RNA is the first step for a variety of gene expression analyses. Very often, complementary studies at the protein level are also required. Usually these analyses are performed using different aliquots of the same experimental sample. However, when working with rare, difficult to obtain, or very small samples, it is sometimes impractical to isolate RNA and native proteins independently. In studies involving large numbers of samples, expensive reagents, or inherent variability (e.g., cell transfection), the addition of independent experimental samples is not only costly and time consuming, but may also lead to inconsistent results. To solve these issues, the Protein And RNA Isolation System (PARIS™ system) has been developed. It allows for the isolation of both RNA and protein from the same experimental sample (see Figure). Using the PARIS™ system, RNA and protein can be isolated simultaneously from whole cell lysates. Alternatively, RNA and protein can be isolated from separate nuclear and cytoplasmic fractions. Tissue or cultured cells are first homogenized in ice-cold Cell Disruption Buffer to prepare a total cell lysate. Since the homogenization is performed quickly on ice and in the presence of detergent, both protein and RNA can be purified directly from this lysate. For RNA isolation, a part of the total cell lysate is immediately mixed with an equal volume of Lysis/Binding Solution. Total RNA is then purified from the mixture using an RNA binding glass fiber filter. After three rapid washing steps, high-quality RNA is eluted in a concentrated form. The entire procedure can be completed in less than 20 minutes. Note: This kit is not recommended for tissues with high levels of ribonucleases, such as pancreas.
Compatible with Most Downstream Applications
The RNA isolated from total, nuclear, or cytoplasmic fractions with the PARIS™ procedure can be used in a variety of downstream applications, including blot hybridization, in vitro translation, cDNA synthesis, and RT-PCR. A DNase I treatment is recommended for RNA that will be used for RT-PCR experiments (see Accessory Products). Protein fractions can be used directly for most common applications, including functional assays, immunoprecipitation, western blotting, or two-dimensional gel electrophoresis (see Figures).
Accessory Products:
TURBO™ DNA-free™ or DNA-free™ DNase Treatment and Removal Reagents (Cat. No. AM1907 and AM1906, respectively) are ideal to quickly remove trace amounts of DNA from the total and nuclear RNA samples without phenol extraction or alcohol precipitation. The cytoplasmic RNA fraction is virtually free of DNA contamination.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | • 30 mL Cell Disruption Buffer; 4°C • 25 mL Cell Fractionation Buffer; 4°C • 30 mL 2X Lysis/Binding Solution; 4°C • 40 mL Wash Solution 1; 4°C • 80 mL Wash Solution 2/3 Concentrate; 4°C • 4 mL Lithium Chloride Precipitation Solution; 4°C • 50 Filter Cartridges; room temperature • 100 Collection Tubes; room temperature • 10 mL Elution Solution; room temperature |
| Isolation Technology | Spin Column (Glass Fiber Filter) |
| Purification Time | 20 min. |
| Elution Volume | 50 to 200 μL |
| Sample Type | Cells, Tissue |
| Final Product Type | Total RNA and Protein |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| For Use With (Application) | RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern/Western blotting, in vitro translation, nuclease protection assays, nucleic acid labeling, enzymatic assays, immunoprecipitation, gel shift assays, 2D gel electrophoresis |
| Quantity | 50 Preps |
| Shipping Condition | Room Temperature |
| Show More |
Frequently Asked Questions (FAQs)
RNA and/or protein can be isolated from either whole cells or from nuclear and cytoplasmid cell partitions.
Quantitation of protein samples obtained with the PARIS kit (Cat. No. AM1921) can be assessed with standard protein assays like BCA, Bradford, or Lowry. From our Protein Assays portfolio, the following assays can be performed in microplates, require only 10 µL of sample, and show high linearity:
-Pierce 660nm Protein Assay Kit (Cat. No. 22662): working range of 50–2000 µg/mL, performed at room temperature, 5-min incubation time, and signal read at 660 nm.
-Pierce Microplate BCA Protein Assay Kit - Reducing Agent Compatible (Cat. No. 23252): working range of 125-2000 µg/mL, performed at 37 degrees C, 50 min incubation, and signal read at 562 nm.
-Pierce Rapid Gold BCA Protein Assay Kit (Cat. No. A53225): working range of 125-2000 µg/mL, performed at room temperature, 5 min incubation, and signal read at 480 nm.
To identify, compare, and choose the best protein assay for your sample type and application, you can use our interactive Protein Assay Selection Guide.
For Research Use Only. Not for use in diagnostic procedures.