Learn More
Invitrogen™ NorthernMax™ Prehybridization/Hybridization Buffer
Description
- Functionally tested by northern blotting and is certified Rnase-free
- Contains 50% formamide with proprietary blocking and background reduction agents
- Blocking agents minimize cross hybridization to ribosomal sequences
- Signal-to-noise ratios are increased by the background reducing agents' suppression of hybridization to the membrane, which is particularly important with nonisotopic detection protocols
- Same component included in the NorthernMax Ultrasensitive Northern Blotting Kits but in a larger size
- Ready to use
- Comprehensive instructions are included
DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Store at 4°C. |
| Concentration | 1X |
| Quantity | 500 mL |
| Shipping Condition | Room Temperature |
| Product Type | Northern Analysis Prehybridization/Hybridization Buffer |
Frequently Asked Questions (FAQs)
Incomplete transfer is often caused by short-circuiting. Strips of Parafilm sealing film around the outside edges of the gel can prevent this. Large RNA species may not transfer well because of their size. A basic transfer buffer (e.g., NorthernMax One-Hour Transfer Buffer) will partially shear the RNA so that larger RNA species transfer more efficiently. Check RNA transfer by including ethidium bromide in RNA samples or staining the gel in ethidium bromide after transfer and viewing your gel under UV light. RNA markers are invaluable to demonstrate whether large RNAs have fully transferred. Our Invitrogen Millennium Markers are especially useful for this purpose, since they include transcripts at 1,000 nt intervals from 0.5 to 9 kb.
The following reasons could have led to cross-hybridization:
There are several types of background, and each can have a different cause:
1) Blotchy signal across the membrane:
This can be caused by a membrane of poor quality, one that has dried out, or one that has been mishandled (e.g., oil from human skin, powder from gloves). Use high quality nylon membrane that has not previously been handled and use forceps to handle the membrane from the edges. Blotchiness can also be caused by uneven distribution of the hybridization reagents. Do not pipette probe directly onto the membrane in hybridization solution; dilute it into the hybridization solution first.
2) A smear through the lane:
Hybridization conditions that are substantially below the optimum for a given probe can lead to high lane-specific background and/or substantial cross-hybridization. Start with a high hybridization temperature and slowly decrease the temperature until a specific signal is obtained. High probe concentrations, especially for nonisotopic probes, can also cause lane-specific background. Use 10 pM nonisotopically labeled DNA probes and 0.1 nM nonisotopically labeled RNA probes.
3) Speckling across the membrane:
Probe preparations with poor incorporation (or where unincorporated nucleotides have not been removed) can cause speckling on the membrane. Check probe quality and remove unincorporated nucleotides. Particulates in probe preparations or hybridization buffer (e.g., when not completely in solution) can also cause speckling on the membrane. Ensure that these reagents are in solution, and consider centrifuging in a microfuge or low-speed centrifuge, or filtering the solutions through a 0.22 µm filter to remove particulates.
If you see high background that is not associated with the lanes, this could be due to:
Poor signal could be a result of the following:
- A hybridization temperature that was not optimal.
- Probe degradation (too old).
- A low specific activity probe (should be ~2 x 10^9 cpm/ug, random primed).
- A probe that was not denatured (DNA).
- A probe concentration that was too low (<10^6 cpm/mL).
- A longer hybridization time needed.
- Poor transfer of RNA to membrane.
- Inadequate cross-linking or overexposure to UV light.
- An alkaline transfer time that was too long (>4 hours).
- The wrong membrane (nitrocellulose).
- Failure to follow nonisotopic detection protocols.
- A message that co-migrates with ribosomal RNA.
- Inappropriate use of intensifying screens.
Residual RNA could be due to:
- An inadequate volume of transfer buffer (>0.5 mL/cm2)
- Too much weight
- The gel used was too thick (>6 mm)
- The transfer time was insufficient (we recommend 15 min per mm of gel thickness)
For Research Use Only. Not for use in diagnostic procedures.