missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MICROBEnrich™ Kit

Catalog No. AM1901
Encompass
Change view
Click to view available options
Quantity:
20 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM1901 20 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. AM1901 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' AM1901
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

For the separation and isolation of bacterial RNA from mixed samples.

For the separation and isolation of bacterial RNA from mixed samples. The Ambion™ kit includes sufficient reagents for depleting 500 μg of host cell RNA (20 reactions x 25 μg RNA) from a complex host-bacterial RNA mixture. Gene expression analysis following host-bacterial interactions is revolutionizing our understanding of the host response to bacterial infection. Numerous studies have utilized microarray analysis to follow host cell transcriptome alterations in response to interactions with bacteria. However, because no method has existed to isolate bacterial RNA away from host cell RNA, the corresponding analyses of bacterial transcriptomes has been impossible. The MICROBEnrich™ Kit enables whole genome microarray expression analyses of bacterial RNA after host-bacterial interactions.

Start With Any Total RNA Sample
For microarray analysis and other sensitive applications, a DNase treatment is recommended to remove contaminating genomic DNA from the total RNA prior to the MICROBEnrich™ Kit procedure.

Remove >90% of Mammalian RNA from Complex Total RNA Mixtures
The MICROBEnrichKituses hybridization capture technology (patent pending) to remove human, mouse, and rat RNA (both mRNA and rRNA) from complex host-bacterial RNA populations, leaving behind enriched microbial total RNA. In the first step of the procedure, host-bacterial total RNA is incubated with a mixture of capture oligonucleotides that bind the mammalian 18S and 28S rRNAs and polyadenylated RNAs. Next, the rRNA/oligo nucleotide hybrids and all polyadenylated mRNAs are removed from the mixture with oligonucleotide-derivatized magnetic beads. After magnetic separation, the bacterial RNA remaining in the supernatant and can be precipitated with ethanol. An Agilent 2100 bioanalyzer electropherogram (see Figure) shows nearly complete removal (>95%) of the host 18S and 28S rRNAs from a mixed sample. Bacterial mRNAs purified with the MICROBEnrich™ Kit are better templates than total RNA for downstream applications and therefore deliver dramatic increases in sensitivity when used in applications such as array analysis (see Figure).

The MICROBEnrich™ Kit Will Work With Any Bacterial Species
The MICROBEnrich Kit was optimized using E. coli RNA in a background of human, mouse, and rat RNA. However, the kit should perform well with RNA from any bacterial species. Although many mammalian host species may be compatible with the MICROBEnrichKit, only human, mouse, and rat rRNA sequences were used for the design and optimization of the capture oligonucleotides.

Specifications

Content And Storage Store Control RNA, Capture Oligo Mix, Glycogen, and 3M NaOAc at -20°C.

Store Oligo MagBeads, Binding Buffer, Wash Solution, Regeneration Solution 1, Regeneration Solution 2, and Resuspension Solution at 4°C.

Store Nuclease-free Water, 1.5 mL tubes and 2 mL collection tubes at room temperature.
Isolation Technology Hybridization Capture, Magnetic Bead
Sample Type Bacteria, RNA, Total RNA (Mammalian Host-Bacterial RNA Mixture)
Final Product Type Total RNA (Bacterial)
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Microarray Analysis
No. of Reactions 20 Preps
Product Line Ambion, MICROBEnrich
Quantity 20 preps
Target Total RNA
I am using the MICROBEnrich Kit to separate bacterial RNA from mouse RNA but am having problems with RNA degradation. Can you offer some tips?

Here are possible causes and solutions:

1. Input total RNA is degraded: to see whether the RNA used in the MICROBEnrich procedure was degraded from the start, evaluate a 0.5-5 µg sample of the input RNA mixture on a denaturing agarose gel (see section III.C starting on page 15 of the manual [https://tools.thermofisher.com/content/sfs/manuals/cms_057050.pdf]). Total RNA should produce rRNA bands that appear sharp and well-defined (Figure 3 on page 16). In high-quality RNA samples, the 28S rRNA band will be 1.5-2 fold brighter than the 18S rRNA band.
2. Practice RNase-free technique: all of the typical precautions against RNase contamination should be observed. Gloves should be worn at all times and changed frequently to avoid the introduction of RNases. Bags containing the centrifuge tubes and the solution tubes and bottles should be kept closed when they are not in use to avoid contamination with dust. Any tubes or solutions that will contact the RNA (that are not supplied with the kit) should be free of RNases.
3. Degradation caused by the heat denaturation step: RNA degradation can occur at elevated temperatures when there are divalent cations in the solution. If your RNA solution does not contain at least 1 mM EDTA, contaminating divalent cations could cause RNA hydrolysis during the heat denaturation step (step II.B.3 on page 8).

For Research Use Only. Not for use in diagnostic procedures.