missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagMAX™-96 Blood RNA Isolation Kit

Catalog No. AM1837
Encompass
Change view
Click to view available options
Quantity:
96 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM1837 96 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM1837 Supplier Invitrogen™ Supplier No. AM1837

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This kit is for the rapid, high-throughput purification of pathogen RNA from mammalian whole blood, milk, and other biofluids in 96-well plates.

This Ambion™ kit is for the rapid, high-throughput purification of pathogen RNA from mammalian whole blood, milk, and other biofluids in 96-well plates. RNA purified using the kit is ideal for RT-PCR applications. The kit uses magnetic bead–based purification and includes sufficient reagents for 96 reactions.

• Easily isolate viral or total RNA from whole blood
• Save precious time—complete the protocol in less than 1 hour
• Walk away—integrate with established robotic platforms
• Validated with human, bovine, porcine, and ovine whole blood

Benefits of MagMAX™ Magnetic Bead-Based Purification:
Magnetic beads offer many benefits compared to other technologies for isolating RNA. Beads bind RNA more efficiently than glass fiber filters, resulting in higher and more consistent RNA yields. Additionally, because filters are not used, there is no risk of filter clogging due to cellular particulates in samples. Since only a small volume of magnetic beads is needed for high- efficiency binding, the bound RNA can be eluted in just 30–50 μL of nuclease-free water, concentrating RNA from large, dilute samples.

Isolate RNA from as Little as 50 μl of Blood:
For the highest-quality RNA, use whole blood samples that have not been frozen. Approximately 300–500 ng of intact RNA can be consistently purified from 50 μl of human whole blood. Bovine, porcine, and ovine whole blood samples can also be used. Yields vary from species to species (see data). The recovered RNA is devoid of PCR inhibitors such as proteins and heme, commonly found in RNA isolated from blood.

Isolate Viral and Total RNA from Whole Blood Samples:
The MagMAX™-96 Blood RNA Isolation Kit eliminates sample preprocessing, which can cause sample mix-up and cross-contamination. The procedure is fast (less than 1 hour), simple, and well suited for automation. The kit can also be used to process fewer than 96 samples at one time and is compatible with either manual processing using multi-channel pipettors or processing with robotic liquid handlers. Downloadable protocols for the use of this kit with specific liquid handling systems are available at our Automation Resource. The MagMAX™-96 Blood RNA Isolation Kit can also be used for total nucleic acid isolation by omitting the DNase treatment.

Specifications

Content And Storage Store Processing Plate with Lid, Lysis/Binding Solution Concentrate, Wash Solution 1 Concentrate, Wash Solution 2 Concentrate, and Elution Buffer at room temp.

Store MagMax Turbo DNase Buffer and RNA Binding Beads at 4°C.

Store Lysis/Binding Enhancer and TURBO DNase at -20°C.
Format 96-well Plate
Isolation Technology Magnetic Bead
Sample Type Liquid Samples (e.g. Serum), Milk, Viral Samples, Whole Blood (Mammalian)
Final Product Type Total RNA, Viral RNA
High-throughput Compatibility High-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
For Use With (Equipment) MagMAX™ Express 24 and 96, Robotic Liquid Handling Systems, MagMAX™ Express-96 Standard Magnetic Particle Processor
For Use With (Application) Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Northern Blotting
No. of Reactions 96 Preps
Prep Scale < 100 μL Blood Samples (Small-scale)
Product Line Ambion, MagMAX
Quantity 96 preps
Starting Material Amount Up to 50 μl whole blood
Target Viral DNA/RNA, Total RNA
Yield 0.5 μg
Show More Show Less
Are MagMax kits compatible with RNAlater reagent-treated samples?

Only MagMax96 for Microarrays Total RNA Isolation Kit (Cat. No. AM1839) is compatible, since it uses Trizol reagent and most of the salt carry-over from RNAlater reagent is eliminated during the lysis steps and partitioned from the aqueous RNA layer on top. For other MagMax kits, optimization is needed. High salt carry over is not optimal for MagMax RNA isolation steps.

Can I edit the manufactured KingFisher Flex protocols?

No, manufactured protocols are locked for editing, but they can be renamed using the ‘save as' function and edited as a new version.

Can I use magnetic beads from other vendors with the KingFisher Flex instrument?

For nucleic acid extraction, we recommend using our MagMAX kits and MagJET kits. Other magnetic beads should work well as long as they are 0.5 to 10 µm in size. However, you will have to write your own KingFisher protocols using the Thermo Scientific BindIt software and validate/optimize the protocol on your own.

I am working with liquid samples with high volume and want to extract nucleic acid from them using the KingFisher Flex instrument. Can I use more volume than the protocol advises?

Yes, this can be done by sample splitting. Set the protocol so that the magnet will collect samples from multiple wells before continuing to the next step. Be sure not to overload.

Can I scale up my reaction volume with the KingFisher Flex instrument?

Yes, reaction volumes can be scaled up, and all other volumes (i.e. wash volumes) will have to be increased by the same fold difference. It is generally not recommended to scale up reaction volumes more than 2-fold unless the plate's volume capacity will allow it.

Are all of the Applied Biosystems MagMAX kits validated for the KingFisher Flex Magnetic Particle Processor?

Yes, all MagMAX kits can be used with the KingFisher Flex Magnetic Particle Processor.


For Research Use Only. Not for use in diagnostic procedures.