missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LeukoLOCK™ Total RNA Isolation System

Catalog No. AM1923
Encompass
Change view
Click to view available options
Quantity:
20 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM1923 20 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM1923 Supplier Invitrogen™ Supplier No. AM1923

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The LeukoLOCK™ Total RNA Isolation System is an innovative method for cellular fractionation of whole blood and total RNA stabilization and extraction from the leukocyte population.

The LeukoLOCK™ Total RNA Isolation System is an innovative method for cellular fractionation of whole blood and total RNA stabilization and extraction from the leukocyte population. The kit is comprised of a LeukoLOCK Fractionation & Stabilization Kit (also available separately; Cat. No. AM1933) and a LeukoLOCK Total RNA Isolation Kit.

The LeukoLOCK system is optimized for use with human blood. Blood is a storehouse of cellular information; however, the presence of globin mRNA in RNA prepared from whole blood can interfere with downstream expression profiling applications. The LeukoLOCK system employs filter-based leukocyte-depletion technology to isolate leukocytes from whole blood and RNAlater™ solution to stabilize the cells on the filter. By excluding red blood cells, the RNA that is purified from captured leukocytes is inherently depleted of globin mRNA, which improves sample utility for expression profiling and other applications.

Leukocyte Capture and Stabilization
A 9–10 mL sample of anticoagulated blood is passed through a LeukoLOCK filter, which captures the total leukocyte population while eliminating red blood cells (including reticulocytes), platelets, and plasma. After rinsing with phosphate-buffered saline, the filter is flushed with RNAlater solution to stabilize the RNA in the captured leukocytes . The RNA can be isolated immediately, or stabilized cells can be maintained for several days at room temperature or for longer periods at –20°C or –80°C.

RNA Isolation
Purification of the RNA is done by first disrupting the captured cells in a guanidinium thiocyanate–based solution that releases the RNA while simultaneously inactivating nucleases. The cell lysate is collected and briefly treated with Proteinase K. RNA is then purified from the lysate using MagMAX magnetic bead-based technology for washing, followed by DNase treatment (with TURBO DNase™) and final clean-up.

Expression Studies
Both quantitative RT-PCR and microarray expression studies have validated the suitability of the purified RNA for reliable transcriptome profiling. Compared to RNA processed with a commonly used blood RNA isolation procedure, RNA isolated with the LeukoLOCK system produces longer median aRNA after amplification and higher percent present calls without requiring additional post-extraction steps to remove globin mRNA. Since globin aRNA represents 25–40% of the aRNA peak from whole blood total RNA, the yield of total RNA is understandably lower with methods that deplete unwanted globin transcripts. RNA amplified after LeukoLOCK purification offers greater array sensitivity due to the dramatic reduction in competing globin transcripts.

Kit Yields
Recovery varies from donor to donor, but is typically 10–20 μg of RNA per 9–10mL of whole blood. RNA recovered using the LeukoLOCK system contains less than one-tenth the amount of reticulocyte-derived alpha- and beta-globin mRNAs present in typical RNA samples from unfractionated whole blood.

  • Fractionates leukocytes from whole blood in minutes
  • Extracts total RNA from leukocytes
  • No additional globin reduction steps required
  • Stabilizes RNA to preserve expression profile
  • Preserves leukocytes at room temperature for days

Not high-throughput compatible (Manual)

Specifications

Content And Storage Store 1X PBS pH 7.4, RNAlater;™ Solution, Rubber Septum Cap, White Slip Connector, Transfer Spike, LeukoLOCK™ Filter, Elution Solution, LeukoLOCK™ Lysis/Binding Solution, LeukoLOCK™ pH Adjustment Solution, Processing Tubes, Wash Solution 1 Concentrate, Wash Solution 2/3 Concentrate and Nuclease-Free Water at room temperature.

Store RNA Binding Beads at 4°C.

Store 1X LeukoLOCK™ DNase Buffer, Proteinase K, and TURBO™ DNase at -20°C.
Isolation Technology Leukocyte Capture Filter, Magnetic Bead
Sample Type Cells, Blood
Final Product Type Total RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Microarray Analysis, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Nuclease Protection Assays, Northern Blotting, cDNA Library Construction
No. of Reactions 20 Preps
Prep Scale > 1 mL Blood Samples (Large-scale)
Product Line Ambion, LeukoLOCK
Quantity 20 preps
Starting Material Amount Up to 10 ml whole blood
Target Total RNA
Show More Show Less
Can I isolate total RNA (including miRNA) using the LeukoLOCK Total RNA Isolation System?

Yes. You will need to follow the alternate protocol, available in the following link.

Can I use heparin for blood collection instead of EDTA when using the LeukoLOCK Total RNA Isolation System?

We have processed blood stored with heparin with the LeukoLOCK Total RNA Isolation System. The RNA looks fine. But we have not tested it for any downstream applications. We discourage your use of heparin, as heparin notoriously inhibits downstream applications such as RT-PCR.

Is the LeukoLOCK Total RNA Isolation System compatible with PAXgene tubes?

No, they are based on different chemistries.

Can I use the LeukoLOCK Total RNA Isolation System for monkey or rodent blood?

The LeukoLOCK Total RNA Isolation System has been tested only for human blood.

Can I use the LeukoLOCK Total RNA Isolation System with less than 3 mL of human blood?

No. The kit has not been validated for less than 3 mL of human blood. It is best to use 9-10 mL blood.

What is the typical RNA yield and quality when using the LeukoLOCK Total RNA Isolation System?

RNA yield is typically 10-20 µg from 10 mL of blood. RNA quality depends on the quality of the blood. Typically, the RNA integrity number (RIN) value is around 9, and the 28S/18S ratio is around 1.5.

What are the advantages of using the LeukoLOCK Total RNA Isolation System?

RNA recovered using the LeukoLOCK Total RNA Isolation System typically contains less than 10% of the amount of reticulocyte-derived alpha- and beta-globin mRNAs present in typical mRNA samples from unfractionated whole blood. The isolated RNA is specially suited for gene expression profiling, as it contains a lower proportion of globin mRNA. We sell LeukoLOCK Fractionation and Stabilization Kit (Cat. No. AM1933) for blood collection and stabilization. You can then use LeukoLOCK Total RNA Isolation Kit (Cat. No. AM1934) to isolate RNA stabilized on the LeukoLOCK filters. Cat. Nos. AM1934 and AM1933 are available together, packaged in a single kit as the LeukoLOCK Total RNA Isolation System (Cat. No. AM1923).


For Research Use Only. Not for use in diagnostic procedures.