missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mirVana™ PARIS™ RNA and Native Protein Purification Kit

Catalog No. AM1556
Encompass
Change view
Click to view available options
Quantity:
40 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM1556 40 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM1556 Supplier Invitrogen™ Supplier No. AM1556

Includes

30mL miRNA Wash Solution I; 50mL Wash Solution 2/3; 80 Collection Tubes; 40 Filter Cartridges; 25mL Cell Disruption Buffer; 25mL 2X Denaturing Solution; 2 x 25mL Acid-Phenol:Chloroform; 5mL Elution Solution

The mirVana™ PARIS™ Kit is for the purification of both native protein and RNA (including small RNA) from the same sample.

The mirVana™ PARIS™ Kit is for the purification of both native protein and RNA (including small RNA) from the same sample. Each kit contains sufficient reagents and consumables for either 40 isolations of protein and total RNA (including small RNAs), or 20 isolations of protein and separate large and small RNA fractions.

How the kit works
The mirVana PARIS Kit procedure begins with homogenization of samples with a special cell disruption buffer that includes nonionic detergent. Protein remains intact, so a portion of the lysate can be used directly for common protein analysis applications. The remainder of the lysate is used for RNA isolation, using a procedure that combines the advantages of organic and solid-phase extraction, while avoiding the disadvantages of both.

  • Isolate small RNA-containing total RNA and native protein from the same sample
  • Start with 100–107 cultured cells or up to 100 mg of many mammalian tissues
  • Enrich for small RNA (<200 nt) to increase sensitivity in downstream analyses
  • Follow a simple, 30-minute procedure
  • Use to correlate mRNA, miRNA or siRNA, and protein levels

Not high-throughput compatible (Manual)

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage • 30 mL miRNA Wash Solution I (room temperature)
• 50 mL Wash Solution 2/3 (room temperature)
• 80 Collection Tubes (room temperature)
• 40 Filter Cartridges (room temperature)
• 25 mL Cell Disruption Buffer (4°C)
• 25 mL 2X Denaturing Solution (4°C)
• 2 × 25 mL Acid-Phenol:Chloroform (4°C)
• 5 mL Elution Solution (-20°C, 4°C, or room temperature)
Isolation Technology Organic Extraction, Cell Lysis (Nonionic Detergent), Spin Column (Glass Fiber Filter)
Purification Time 30 min.
Sample Type Cell Cultures, Tissue (Mammalian)
Final Product Type Total RNA, Transcriptome RNA, siRNA, Protein (Native), snRNA, micro RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Western Blotting, microRNA Analysis, Northern Blotting, cDNA Library Construction
No. of Reactions 40 Preps
Product Line mirVana, Ambion
Quantity 40 preps
Shipping Condition Room Temperature
Starting Material Amount Up to 10^7 cells, Up to 100 mg tissue
Target Total RNA, RNA and Protein, siRNA, miRNA
Show More Show Less
I'd like to isolate RNA and protein from the same sample. Do you have a kit for this?

Yes, you can use TRIzol Reagent or the mirVana PARIS RNA and Native Protein Purification Kit.

For Research Use Only. Not for use in diagnostic procedures.