missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Glycogen (5 mg/ml)

Catalog No. AM9510
Encompass
Change view
Click to view available options
Quantity:
5 tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9510 5 tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM9510 Supplier Invitrogen™ Supplier No. AM9510

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Glycogen is a branched chain carbohydrate and is useful as a nucleic acid coprecipitant.

Glycogen is a branched chain carbohydrate and is useful as a nucleic acid coprecipitant. It is provided in five tubes of 1 mL each at a concentration of 5 mg/mL.

  • Ideal for RT-PCR
  • Increases pellet mass
  • Quantitative recovery of low concentrations (ng/mL) of nucleic acid
  • Prevents pellet loss in nuclease protection assays

What is a coprecipitant?
Co-precipitants are inert substances used to aid recovery of nucleic acids during alcohol precipitations. While they can be used for precipitating large amounts of nucleic acids, they are essential for quantitative recovery of small amounts of nucleic acids in dilute solutions. Often, the use of such molecules is desirable for no other reason but visualization of the pelleted precipitate after centrifugation. When used at a final concentration of 50 to 150 μg/mL, glycogen will coprecipitate with nucleic acids in the presence of 0.5M ammonium acetate and isopropanol or ethanol. It is supplied at a concentration of 5mg/mL in diethyl pyrocarbonate-treated distilled water. Glycogen will not interfere with 260/280 readings. This glycogen product is isolated from mussel, a biological source, as are most other preparations of this coprecipitant. Glycogen is treated with Proteinase K and SDS to remove any contaminating nucleases, and then phenol/chloroform extracted, ethanol precipitated, and resuspended in nuclease-free water. The glycogen is guaranteed RNase- and DNase-free.

Order Info

Shipping Condition: Dry Ice

Specifications

Concentration 5 mg/mL
Content And Storage Store at –20°C.
For Use With (Application) RNA/DNA Extraction
Product Line Ambion
Product Type Glycogen
Purity Molecular Biology Grade
Quantity 5 tubes
Shipping Condition Dry Ice
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.