missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GlycoBlue™ Coprecipitant (15 mg/mL)
Description
GlycoBlue Coprecipitant consists of a blue dye covalently linked to glycogen, a branched chain carbohydrate, which is useful as a nucleic acid coprecipitant. The attached dye increases visibility of the pellet. This product is an ideal coprecipitant in nuclease protection assays at 1/100 dilution of stock solution.
Features of GlycoBlue™ Coprecipitant:
• Ideal for RT-PCR
• Increases pellet mass and visibility
• Quantitative recovery of low concentrations (ng/mL) of nucleic acid
• Prevents pellet loss in nuclease protection assays
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
| Concentration | 15 mg/mL |
| Content And Storage | Store at –20°C. |
| Packaging Type | Tube |
| Product Line | Ambion, GlycoBlue |
| Purity | Molecular Biology Grade |
| Quantity | 5 x 300 μL |
| Shipping Condition | Dry Ice |
| Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.