Learn More
Invitrogen™ DEPC-Treated Water DFS Item
Ambion DEPC-treated water is certified nuclease-free. DEPC-treated water is autoclaved pre- and post-packaging to ensure sterility and inactivation of DEPC.
Supplier:  Invitrogen™ AM9920
 View more versions of this product
Catalog No. AM9920
Only null left
Explore available promotions
Description
Description
Ambion DEPC-treated water is certified nuclease-free. DEPC-treated water is autoclaved pre- and post-packaging to ensure sterility and inactivation of DEPC. It is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Specifications
Specifications
6 to 8
Bottle
1 x 500 mL
Room Temperature
Molecular Biology
DEPC-Treated
Autoclaved, Membrane-Filtered
Ambion
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.