missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DEPC-Treated Water

Catalog No. p-4926893
Encompass
Change view
Click to view available options
Quantity:
10 x 50 mL
1 x 100 mL
1 x 500 mL
5 x 100 mL
1 x 1000 mL
4 x 1000 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9915G 1 x 100 mL
AM9920 1 x 500 mL
AM9906 10 x 50 mL
AM9916 5 x 100 mL
AM9922 1 x 1000 mL
4387937 4 x 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. AM9915G Supplier Invitrogen™ Supplier No. AM9915G
Only null left

Ambion DEPC-treated water is certified nuclease-free. DEPC-treated water is autoclaved pre- and post-packaging to ensure sterility and inactivation of DEPC.

Ambion DEPC-treated water is certified nuclease-free. DEPC-treated water is autoclaved pre- and post-packaging to ensure sterility and inactivation of DEPC. It is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Specifications

Grade Molecular Biology
Packaging Bottle
Treatment(s) DEPC-Treated
Quantity 1 x 100 mL
Purification Method Autoclaved, Membrane-Filtered
Shipping Condition Room Temperature
Product Line Ambion

For Research Use Only. Not for use in diagnostic procedures.