missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Buffer Kit, RNase-free

Catalog No. AM9010
Encompass
Change view
Click to view available options
Quantity:
8 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9010 8 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM9010 Supplier Invitrogen™ Supplier No. AM9010
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Invitrogen Buffer Kit provides high quality, RNase free buffers for critical RNA analysis procedures.

The Invitrogen Buffer Kit provides high quality, RNase free buffers for critical RNA analysis procedures. The kit consists of one 100 mL bottle of each of the following solutions: 1M Tris pH 7.0, 1M Tris pH 8.0, 5M NaCl, 1M MgCl2, 0.5M EDTA pH 8.0, 2M KCl, 5M Ammonium Acetate, and DEPC-treated H2O. Ammonium Acetate is sterile filtered. All other solutions are autoclaved post-packaging to ensure sterility. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

Specifications

For Use With (Application) miRNA analysis
Physical Form Liquid
Product Line Ambion
Quantity 8 x 100 mL
DNase DNase-Free, RNase-Free

For Research Use Only. Not for use in diagnostic procedures.