missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BSA, acetylated (20 mg/mL)

Catalog No. AM2614
Encompass
Change view
Click to view available options
Quantity:
6 x 25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM2614 6 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM2614 Supplier Invitrogen™ Supplier No. AM2614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Invitrogen Acetylated BSA is used as a carrier protein and as an enzyme stabilizer in which the absence of contaminants such as nucleases and proteases is essential.

Invitrogen Acetylated BSA is used as a carrier protein and as an enzyme stabilizer in which the absence of contaminants such as nucleases and proteases is essential. Six tubes of 25mg are provided, at a concentration of 40 to 50mg/mL. The acetylation process inactivates any trace nuclease activity that might be present. While the acetylation process does change BSA's binding characteristics and makes it inhibitory in PCR reactions, it is routinely added to restriction digests. The molecular weight is ∽66 kd. Acetylated BSA is rigorously tested for contaminating nonspecific endonuclease, exonuclease, protease, and RNase activity.

Order Info

Shipping Condition: Dry Ice

Specifications

Concentration 20 mg/mL
Physical Form Liquid
Product Line Ambion
Quantity 6 x 25 mg
Packaging Tube

For Research Use Only. Not for use in diagnostic procedures.