missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BrightStar™-Plus Positively Charged Nylon Membrane, 15 cm x 15 cm

Catalog No. AM10100
Encompass
Change view
Click to view available options
Quantity:
5 pieces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM10100 5 pieces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM10100 Supplier Invitrogen™ Supplier No. AM10100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with radiolabeled and nonisotopic probes for northern and other blot applications

  • For consistent, reliable results with minimal background
  • Five sheets measuring 15 x 15cm are provided
  • This membrane exhibits exceptionally low nonspecific binding in nonisotopic detection applications when used with proprietary blocking and wash solutions

DNA and RNA Purification and Analysis, Dot Blotting, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Proteins, Expression, Isolation and Analysis, Southern Blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store at room temperature.
Membrane Nylon Membrane
Quantity 5 pieces
Shipping Condition Room Temperature
Product Type Nylon Membrane

For Research Use Only. Not for use in diagnostic procedures.