missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Amberlite™ IRA402, Chloride Form
Strongly basic
$374.14 - $374.14
Specifications
| pH | 0 to 14 |
|---|---|
| Product Line | Amberlite |
| Capacity (Metric) | 1.2 meq/mL by wetted bed volume |
| MDL Number | MFCD00145564 |
| CAS | 52439-77-7 |
Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow