missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Amberlite™ IRA402, Chloride Form

Catalog No. 111014455
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014455 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014455 Supplier MilliporeSigma™ Supelco™ Supplier No. 064661KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Strongly basic

Specifications

Particle Size 500 to 1000 μm
Technique Chromatography
pH 0 to 14
Quantity 1 kg
Product Line Amberlite
Type Anion Exchange Resin
Material Styrene-divinylbenzene
Capacity (Metric) 1.2 meq/mL by wetted bed volume
MDL Number MFCD00145564
CAS 52439-77-7

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow