missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Amberlite™ IRA-67, Free Base
Previously known as IRA-68.
$138.12 - $422.77
Specifications
| Product Line | Amberlite |
|---|---|
| Capacity (Metric) | 1.6 meq/mL by wetted bed volume |
| MDL Number | MFCD00145567 |
| CAS | 65899-87-8 |
Description
Amberlite™ IRA 67 is a weakly basic anion exchange resin with high capacity for large organics.Specifications
| Amberlite | |
| 1.6 meq/mL by wetted bed volume | |
| MFCD00145567 | |
| 65899-87-8 |
Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow