missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Amberlite™ IRA-67, Free Base

Catalog No. 111015092
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015092 250 g
111015093 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015092 Supplier MilliporeSigma™ Supelco™ Supplier No. 06411250G

Previously known as IRA-68.

Amberlite™ IRA 67 is a weakly basic anion exchange resin with high capacity for large organics.

Specifications

Quantity 250 g
Product Line Amberlite
Capacity (Metric) 1.6 meq/mL by wetted bed volume
MDL Number MFCD00145567
CAS 65899-87-8

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow