missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AmberChrom™, 50WX8, 100-200 mesh, H-form, Thermo Scientific™
$729.43 - $729.43
Chemical Identifiers
| CAS | 69011-20-7 |
|---|---|
| Molecular Formula | C28H30 |
| Molecular Weight (g/mol) | 0.00 |
| MDL Number | MFCD00132722 |
| InChI Key | NWUYHJFMYQTDRP-UHFFFAOYSA-N |
| IUPAC Name | 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene |
| SMILES | * |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC472925000
|
Thermo Scientific Chemicals
472925000 |
500 g |
Each for $729.43
|
|
|||||
Chemical Identifiers
| 69011-20-7 | |
| 0.00 | |
| NWUYHJFMYQTDRP-UHFFFAOYSA-N | |
| * |
| C28H30 | |
| MFCD00132722 | |
| 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene |
Specifications
| Beads | |
| 42%,50% | |
| ≥1.7 eq/L (H form) | |
| Plastic Bottle | |
| MFCD00132722 | |
| Beige to Green/Beige to Light Brown | |
| Insoluble in water | |
| * | |
| 0.00 |
| 69011-20-7,7732-18-5 | |
| 50%,58% | |
| 100 to 200 mesh, ≤15 % (< 88 μm), ≤15 % (> 250 μm) | |
| C28H30 | |
| 50 to 58 % (H form) | |
| 500 g | |
| NWUYHJFMYQTDRP-UHFFFAOYSA-N | |
| 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene | |
| AmberChrom™ |
Safety and Handling
Corrosion
Recommended Storage : Normal conditions