missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AmberChrom™, 50WX8, 100-200 mesh, H-form, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 472925000
Specifications
| Beads | |
| 69011-20-7,7732-18-5 | |
| 50%,58% | |
| 100 to 200 mesh, ≤15 % (< 88 μm), ≤15 % (> 250 μm) | |
| C28H30 | |
| 50 to 58 % (H form) | |
| 500 g | |
| NWUYHJFMYQTDRP-UHFFFAOYSA-N | |
| 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene |
| AmberChrom™ | |
| 42%,50% | |
| ≥1.7 eq/L (H form) | |
| Plastic Bottle | |
| MFCD00132722 | |
| Beige to Green/Beige to Light Brown | |
| Insoluble in water | |
| * | |
| 0.00 |
Chemical Identifiers
| 69011-20-7 | |
| 0.00 | |
| NWUYHJFMYQTDRP-UHFFFAOYSA-N | |
| * |
| C28H30 | |
| MFCD00132722 | |
| 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene |
Safety and Handling
Corrosion
Recommended Storage : Normal conditions