missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Amber Wide Mouth Packer Glass Bottles with White Polypropylene Caps

Fisherbrand™ Amber Wide Mouth Packer Glass Bottles with White Polypropylene Caps are ideal for storage of light sensitive samples or dry powders.

 View more versions of this product

Catalog No. FB02911451A


Only null left

Description

Description

  • Feature a wide opening for making them the ideal general purpose bottle for solids
  • The amber color filters out ultraviolet wavelengths, making amber packers ideal for light sensitive solids
  • Amber packers are the classic vitamin bottles and are often used in pharmaceutical packaging
  • Easy transfer of contents
  • Good impact strength and pliability
  • Caps can be remelted after they are formed
  • Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
Specifications

Chemical Identifiers

Amber
White
Wide
Pulp/Vinyl
Glass (Type 3)
Yes
Wide Mouth Packer Bottle
Polypropylene

Specifications

500 mL
50 mm
149 mm
60
53-400
Wide
Wide Mouth Packer Bottle
Polypropylene
81 mm
Amber
40 mm
Glass (Type 3)
White
Yes
146 mm
Pulp/Vinyl
53-400 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions