missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Amber Reagent Bottle with Screw Cap

High quality, borosilicate glass flask. Pouring ring for drip free pouring and clean, safe working.

$43.89 - $206.18

Specifications

Color Amber
For Use With (Application) For storing chemicals, liquids or powders
Material Borosilicate Glass
Quantity 1 Ea.
Sterility Non-sterile
View More Specs

Products 5
Catalog Number Mfr. No. Capacity (Metric) Graduations Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Graduations Price Quantity & Availability  
S27986
View Documents Promotion Details
Eisco™
CH0165A
100 mL 10mL
Each for $43.89
Only null left
 
S27987
View Documents Promotion Details
Eisco™
CH0165B
250 mL 50mL
Each for $63.29
Only null left
 
S27988
View Documents Promotion Details
Eisco™
CH0165C
500 mL 50mL
Each for $72.29
Only null left
 
S27989
View Documents Promotion Details
Eisco™
CH0165D
1000 mL 50mL
Each for $138.50
Only null left
 
S27990
View Documents Promotion Details
Eisco™
CH0165E
2000 mL 250mL
Each for $206.18
Only null left
 
Description

Description

  • Reagent bottle made of high quality, amber colored Borosilicate 3.3 glass.
  • Bottle features white graduation lines.
  • As per DIN ISO 4796-1 standard.
  • Equipped with an orange, GL 45 screw cap and features a drip free pouring ring for cleaner, safer transfers of liquids and powders.
  • Both screw cap and pouring ring are autoclavable.
  • Bottle features superior chemical durability and low temperature gradient - can withstand high temperatures and thermal shocks.
Specifications

Specifications

Amber
Borosilicate Glass
Non-sterile
Yes
Yes
Cylindrical
Blank
Autoclavable
For storing chemicals, liquids or powders
1 Ea.
Orange
Reusable
Narrow
Reagent Bottle
Screw Cap
Videos
SDS
Documents

Documents