missing translation for 'onlineSavingsMsg'
Learn More

Amarouciaxanthin A, ≥97% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012279
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012279 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012279 Supplier Merck Emd Millipore Supplier No. 547581MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 100667-78-5
Linear Formula C40H54O5
Type Analytical Standard
Quantity 1 mg
Formula Weight 614.85
Percent Purity ≥97% (HPLC)
Grade Analytical Standard