missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amaranth, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| CAS | 915-67-3 |
| Physical Form | Neat |
| Linear Formula | C20H11N2 Na3O10S3 |
| MDL Number | MFCD00004076 |
| Synonym | Acid Red 27; Azorubin S; FD & C Red Dye No. 2 |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 604.47 |
| Percent Purity | ≥98.0% (HPLC) |
| Show More |