missing translation for 'onlineSavingsMsg'
Learn More

AM 251 97.0+%, TCI America™

Catalog No. p-7118237
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
Catalog No. Quantity
A315110MG 10 mg
A315150MG 50 mg
2 options

Catalog No. A315110MG

Supplier: TCI America A315110MG

Only null left

Chemical Identifiers

CAS 183232-66-8
Molecular Formula C22H21Cl2IN4O
Molecular Weight (g/mol) 555.241
MDL Number MFCD01861181
InChI Key BUZAJRPLUGXRAB-UHFFFAOYSA-N
PubChem CID 2125
IUPAC Name 1-(2,4-dichlorophenyl)-5-(4-iodophenyl)-4-methyl-N-piperidin-1-ylpyrazole-3-carboxamide
SMILES CC1=C(N(N=C1C(=O)NN2CCCCC2)C3=C(C=C(C=C3)Cl)Cl)C4=CC=C(C=C4)I

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 10 mg
Synonym unii-3i4fa44mai, 1-2,4-dichlorophenyl-5-4-iodophenyl-4-methyl-n-piperidin-1-yl-1h-pyrazole-3-carboxamide, n-piperidin-1-yl-5-4-iodophenyl-1-2,4-dichlorophenyl-4-methyl-1h-pyrazole-3-carboxamide, 3i4fa44mai, 1-2,4-dichlorophenyl-5-4-iodophenyl-4-methyl-n-piperidin-1-ylpyrazole-3-carboxamide, n-piperidin-1-yl-5-4-iodophenyl-1-2, 4-dichlorophenyl-4-methyl-1h-pyrazole-3-carboxamide, 1-2,4-dichlorophenyl-5-4-iodophenyl-4-methyl-n-1-piperidinyl-1h-pyrazole-3-carboxamide, am 251 pharmaceutical, tocris-1117, d08yqt
Formula Weight 555.24
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material AM 251
TSCA No
Recommended Storage Freezer