CAS RN 183232-66-8
CAS RN 183232-66-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
AM 251 97.0+%, TCI America™
CAS: 183232-66-8 Molecular Formula: C22H21Cl2IN4O Molecular Weight (g/mol): 555.241 MDL Number: MFCD01861181 InChI Key: BUZAJRPLUGXRAB-UHFFFAOYSA-N PubChem CID: 2125 IUPAC Name: 1-(2,4-dichlorophenyl)-5-(4-iodophenyl)-4-methyl-N-piperidin-1-ylpyrazole-3-carboxamide SMILES: CC1=C(N(N=C1C(=O)NN2CCCCC2)C3=C(C=C(C=C3)Cl)Cl)C4=CC=C(C=C4)I