missing translation for 'onlineSavingsMsg'
Learn More

AM 251 97.0+%, TCI America™

Catalog No. A315110MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A315110MG 10 mg
A315150MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A315110MG Supplier TCI America Supplier No. A315110MG

Chemical Identifiers

CAS 183232-66-8
Molecular Formula C22H21Cl2IN4O
Molecular Weight (g/mol) 555.241
MDL Number MFCD01861181
InChI Key BUZAJRPLUGXRAB-UHFFFAOYSA-N
PubChem CID 2125
IUPAC Name 1-(2,4-dichlorophenyl)-5-(4-iodophenyl)-4-methyl-N-piperidin-1-ylpyrazole-3-carboxamide
SMILES CC1=C(N(N=C1C(=O)NN2CCCCC2)C3=C(C=C(C=C3)Cl)Cl)C4=CC=C(C=C4)I

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 10 mg
Synonym unii-3i4fa44mai, 1-2,4-dichlorophenyl-5-4-iodophenyl-4-methyl-n-piperidin-1-yl-1h-pyrazole-3-carboxamide, n-piperidin-1-yl-5-4-iodophenyl-1-2,4-dichlorophenyl-4-methyl-1h-pyrazole-3-carboxamide, 3i4fa44mai, 1-2,4-dichlorophenyl-5-4-iodophenyl-4-methyl-n-piperidin-1-ylpyrazole-3-carboxamide, n-piperidin-1-yl-5-4-iodophenyl-1-2, 4-dichlorophenyl-4-methyl-1h-pyrazole-3-carboxamide, 1-2,4-dichlorophenyl-5-4-iodophenyl-4-methyl-n-1-piperidinyl-1h-pyrazole-3-carboxamide, am 251 pharmaceutical, tocris-1117, d08yqt
Formula Weight 555.24
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material AM 251
TSCA No
Recommended Storage Freezer