Learn More
Stemgent™ Alvetex™ 3D Cell Growth Plates
A range of product formats for all your applications
$1091.33 - $3176.83
| Catalog Number | Mfr. No. | No. of Wells | Quantity | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Wells | Quantity | Price | Quantity | ||||
AVP0022
|
Stemgent™
AVP0022 |
12 | 2 plates |
Pack for $1,091.33
|
|
||||
AVP0062
|
Stemgent™
AVP0062 |
24 | 2 plates |
Pack for $1,268.43
|
|
||||
AVP0092
|
Stemgent™
AVP0092 |
96 | 2 plates |
Pack for $1,594.13
|
|
||||
AVP00610
|
Stemgent™
AVP00610 |
24 | 10 plates |
Pack for $3,176.83
|
|
||||
Description
Alvetex Scaffold enables genuine 3-dimensional cell culture to be achieved simply and routinely.
- Highly porous (more than 90%) inert polystyrene scaffold
- Provides cultured cells with an environment and physical space in which to grow in 3 dimensions (3D)
- Once seeded, cells freely invade the scaffold and start to produce genuine, homogeneous 3D cellular structures that resemble micro-slabs of tissue
- Plate format (96 well, 24 well and 12 well)
Typical uses:
- Growth of 3D tumor cells for testing cytotoxicity of anti-cancer drugs
- Growth and differentiation of stem cells in 3D
- Improved predictive accuracy of Liver Tox testing with primary hepatocytes
- Formation of full thickness skin models for compound testing
- Growth of complex epithelial models
- Co-culture models
- 3D cell migration/invasion models
- Enhanced cell viability and responsiveness to growth factors/therapeutic agents
- Improve predictive accuracy of cell base assays
- More accurately mirror the natural in-vivo cellular environment
- Better understand how cells interact within complex systems
- Reduce the dependency on animal model testing
- Build more biologically relevant in-vitro assays
Specifications
| 3D Cell Culture Plates and Inserts |