Learn More
United Scientific™ Aluminum Weighing Dishes, Round
Weighing Dishes are ideal for general purpose weighing, dispensing, storing, sampling, and testing applications.
$115.23 - $128.07
Specifications
| Autoclavable | Not Autoclavable |
|---|---|
| Bottom Shape | Flat |
| Disposable | Single-use |
| Shape | Circular |
| Sterility | Non-sterile |
Description
- These smooth-wall, flat-bottom aluminum dishes are ideal for general purpose weighing, dispensing, storing, sampling, and testing applications.
- Dishes include tapered sides to enable easy stacking.
- Ideal for general-purpose applications
- Slightly tapered sides for easy stacking
- Disposable
- Aluminum construction
Specifications
| Not Autoclavable | |
| Single-use | |
| Non-sterile |
| Flat | |
| Circular | |
| Weighing Dish |