missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Aluminum Weighing Boats with Tabs

Dishes for general-purpose weighing, dispensing, storage, food processing, evaporating or drying.

Supplier:  Fisherbrand™ HS14521AF2

 View more versions of this product

Catalog No. 01549002


Only null left

Description

Description

The Aluminum Weighing Boat may be used for general-purpose weighing, dispensing, storage, food processing, evaporating or drying.

Aluminum Weighing Boats:

  • Sturdy yet lightweight boats feature a smooth, flat bottom for sample handling and a flanged, crimped wall for rigidity
  • Finger tab handle can be etched for identification prior to weighing
  • Designed for moisture determination balances with slightly tapered walls for stacking
  • Packaging: 100
Specifications

Chemical Identifiers

Not Autoclavable
Silver
Aluminum
Non-sterile
100/Cs.
Round
Single-use
Circular
Weighing Boat

Specifications

Not Autoclavable
20 mL
Single-use
Circular
Weighing Boat
0.5 in.
43 mm
Round
Silver
Aluminum
Non-sterile
100/Cs.
1.7 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions