missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Aluminum Warning Sign: FLAMMABLE GAS

Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.

Supplier:  Brady™ 131795

 View more versions of this product

Catalog No. 19-106-208


Only null left
Explore available promotions

Description

Description

  • Aluminum material is abrasion-resistant and chemical-resistant
  • Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
Specifications

Specifications

Not Laminated
1 Ea.
Corner Holes
230°F
0.035 in.
Rectangular
10 x 7 in.
Red on White
Aluminum
Non-adhesive
7 in.
-40°F
10 in.
-40°F to 230°F
FLAMMABLE GAS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions