missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Aluminum Warning Sign: BENZENE CANCER HAZARD FLAMMABLE- NO SMOKING AUTHORIZED PERSONNEL ONLY RESPIRATOR REQUIRED
Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.
$19.40 - $35.04
Specifications
| Finish | Not Laminated |
|---|---|
| Material | Aluminum |
| Quantity | 1 Ea. |
| Adhesive Type | Non-adhesive |
| Mounting Type | Corner Holes |
Description
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
| Not Laminated | |
| 1 Ea. | |
| Corner Holes | |
| -40°F | |
| Rectangular | |
| BENZENE CANCER HAZARD FLAMMABLE- NO SMOKING AUTHORIZED PERSONNEL ONLY RESPIRATOR REQUIRED |
| Aluminum | |
| Non-adhesive | |
| 230°F | |
| 0.035 in. | |
| -40°F to 230°F | |
| Red on White |