Learn More
Synthware™ Aluminum Support Jacks
GREENER_CHOICE
Adjustable height jack can be used to support various types of glassware and equipment commonly used in the lab.
$265.39 - $1,004.96
Specifications
Material Aluminum
Product Type Support Jack
For Use With (Equipment) Laboratory Glassware
View More Specs
Products 4
Catalog Number Mfr. No. Dimensions (L x W x H) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W x H) Price Quantity & Availability  
31502288
GREENER_CHOICE
View Documents Promotion Details
Synthware™
J111018
10 x 10 x 14 cm
Each for $265.39
Only null left
31502289
GREENER_CHOICE
View Documents Promotion Details
Synthware™
J111525
15 x 15 x 28 cm
Each for $471.07
Only null left
31502290
GREENER_CHOICE
View Documents Promotion Details
Synthware™
J112028
20 x 20 x 29 cm
Each for $698.75
Only null left
31502291
GREENER_CHOICE
View Documents Promotion Details
Synthware™
J112532
20 x 25 x 38 cm
Each for $1,004.96
Only null left
Description
Description
  • Aluminum support jack
  • Adjustable height
  • Capable of supporting various types of glassware and equipment
Specifications
Specifications
Aluminum
Support Jack
Laboratory Glassware