Learn More
Synthware™ Aluminum Support Jacks
GREENER_CHOICE
Catalog No. 31502288
Change view
Click to view available options
Dimensions (L x W x H):
10 x 10 x 14 cm
15 x 15 x 28 cm
20 x 20 x 29 cm
20 x 25 x 38 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
31502288 10 x 10 x 14 cm
31502289 15 x 15 x 28 cm
31502290 20 x 20 x 29 cm
31502291 20 x 25 x 38 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 31502288 Supplier Synthware™ Supplier No. J111018
Adjustable height jack can be used to support various types of glassware and equipment commonly used in the lab.
  • Aluminum support jack
  • Adjustable height
  • Capable of supporting various types of glassware and equipment
Specifications
Material Aluminum
Dimensions (L x W x H) 10 x 10 x 14 cm
Product Type Support Jack
For Use With (Equipment) Laboratory Glassware