missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Sulfate, Hydrated, Crystal, ACS, 98-102%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 125 g |
| Percent Purity | 98 to 102% |
| Grade | ACS |
| Packaging | Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.