missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Sulfate Hydrate ACS MP Biomedicals
Supplier: MP Biomedicals Inc 0219140391
Specifications
| Aluminum Sulfate Hydrate | |
| 17927-65-0 | |
| 98 to 102% | |
| aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18 | |
| O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3] | |
| 666.401 | |
| CHEBI:74779 | |
| ACS Reagent |
| Hydrate | |
| 1 kg | |
| Al2H36O30S3 | |
| AMVQGJHFDJVOOB-UHFFFAOYSA-H | |
| dialuminum;trisulfate;octadecahydrate | |
| 22377415 | |
| 342.15 (Anhydrous) |
Chemical Identifiers
| 17927-65-0 | |
| aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18 | |
| 22377415 | |
| CHEBI:74779 |
Safety and Handling
Recommended Storage : Store at room temperature.