missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Sulfate Hydrate ACS MP Biomedicals

Catalog No. p-3178620
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19140380 100 g
ICN19140390 500 g
ICN19140391 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19140380 Supplier MP Biomedicals Inc Supplier No. 0219140380
Only null left

Chemical Identifiers

CAS 17927-65-0
MDL Number MFCD00149136
Synonym aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18
PubChem CID 22377415
ChEBI CHEBI:74779

Specifications

Quantity 100 g
Assay Percent Range 98 to 102%
Molecular Formula Al2O12S3
InChI Key DIZPMCHEQGEION-UHFFFAOYSA-H
SMILES [Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
IUPAC Name dialuminium(3+) trisulfate
Molecular Weight (g/mol) 342.13
Formula Weight 342.15 (Anhydrous)
Grade ACS Reagent
Chemical Name or Material Aluminum Sulfate Hydrate, Hydrate
Recommended Storage Store at room temperature.