missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sulfate, anhydrous, 99.99% (metals basis)

Catalog No. p-7059419
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4456306 5 g
AA4456314 25 g
AA4456322 100 g
AA4456336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA4456306 Supplier Thermo Scientific Chemicals Supplier No. 04456306
Only null left

CAS: 10043-01-3 | Al2O12S3 | 342.131 g/mol

Aluminum sulfate acts as a flocculating agent in the purification of drinking water, and in waste water treatment plants. It acts as a mordant in dyeing, printing, textiles and also used in paper manufacturing. It is a waterproofing agent and accelerator in concrete. Further, it is used as a foaming agent in fire fighting foam, photographic film and in photochemicals. It is also used in styptic pencils, pain relief and in dentistry for gingival retraction cords.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon

Chemical Identifiers

CAS 10043-01-3
Molecular Formula Al2O12S3
Molecular Weight (g/mol) 342.131
MDL Number MFCD00003423
InChI Key DIZPMCHEQGEION-UHFFFAOYSA-H
Synonym aluminum sulfate, dialuminum trisulfate, aluminum sulfate 2:3, filter alum, aluminum sulphate, aluminum sulphate, dialuminum sulfate, papermaker's alum, sulfuric acid, aluminum salt 3:2
PubChem CID 24850
ChEBI CHEBI:74768
IUPAC Name dialuminum;trisulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]

Specifications

Melting Point 770°C
pH 3.5
Physical Form Powder
Quantity 5 g
Merck Index 14,366
Solubility Information Soluble in water. Slightly soluble in alcohol and dilute mineral acids. Insoluble in ethanol.
Formula Weight 342.15
Percent Purity 99.99%
Sensitivity Hygroscopic
Density 2.71 g/mL
Assay (metals basis)
Chemical Name or Material Aluminum sulfate, Anhydrous
Show More Show Less
Hazard Category H318
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 233-135-0
RTECSNumber BD1700000
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures

RUO – Research Use Only