missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Sulfate, 18-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5390675
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002255 500 g Poly Bottle
02002254 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02002255 Supplier Avantor Supplier No. 056401
Only null left

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7784-31-8
Molecular Formula Al2H36O30S3
Molecular Weight (g/mol) 666.401
InChI Key AMVQGJHFDJVOOB-UHFFFAOYSA-H
Synonym aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18
PubChem CID 22377415
ChEBI CHEBI:74779
IUPAC Name dialuminum;trisulfate;octadecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]

Specifications

Physical Form Crystals
Quantity 500 g
Formula Weight 666.41
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.69kg
Chemical Name or Material Aluminum Sulfate, 18-Hydrate