missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Aluminum Sign: DANGER - COMPRESSED AIR. DO NOT USE FOR BLOWING OFF CLOTHES.
Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$19.40 - $35.04
Specifications
| Material | Aluminum |
|---|---|
| Shape | Rectangular |
| Displayed Text | DANGER - COMPRESSED AIR. DO NOT USE FOR BLOWING OFF CLOTHES. |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Includes | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Includes | Price | Quantity & Availability | ||||
|
19106434
|
Brady™
144928 |
10 x 7 in. | Caution Picto |
Each for $19.40
|
|
||||
|
19106435
|
Brady™
144929 |
14 x 10 in. | Caution Picto |
Each for $35.04
|
|
||||
|
19107045
|
Brady™
40884 |
14 x 10 in. | Without Picto |
Each for $35.04
|
|
||||
|
19107044
|
Brady™
40883 |
10 x 7 in. | Without Picto |
Each for $19.40
|
|
||||
Description
- Aluminum material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
| Aluminum | |
| Rectangular | |
| DANGER - COMPRESSED AIR. DO NOT USE FOR BLOWING OFF CLOTHES. |